215236
Sulfur
reagent grade, purified by sublimation, -100 mesh particle size, powder
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Empirical Formula (Hill Notation):
S
CAS Number:
Molecular Weight:
32.07
EC Number:
MDL number:
UNSPSC Code:
12141912
PubChem Substance ID:
NACRES:
NA.55
grade:
reagent grade
form:
powder
Recommended Products
grade
reagent grade
Quality Level
vapor density
8.9 (vs air)
vapor pressure
1 mmHg ( 183.8 °C)
10 mmHg ( 246 °C)
form
powder
autoignition temp.
450 °F
purified by
sublimation
resistivity
2E23 μΩ-cm, 20°C
particle size
-100 mesh
bp
444.7 °C (lit.)
mp
112.8 °C (rhombic) (lit.)
117-120 °C (lit.)
119.0 °C (monoclinic) (lit.)
SMILES string
[S]
InChI
1S/S
InChI key
NINIDFKCEFEMDL-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Sulfur is a nonmetallic element that can be used in the preparation of many organosulfur compounds such as 3D sulfur-rich polymers. It can also be used as a mild oxidizing agent for dehydrogenation and Willgerodt reaction.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Irrit. 2
Storage Class Code
4.1B - Flammable solid hazardous materials
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Sulfur
Morrison J
e-EROS Encyclopedia of Reagents for Organic Synthesis (2001)
Electron microscopy study of CVT grown Fe-sulphides.
Zavasnik J and Recnik A.
Journal of Crystal Growth, 367, 18-23 (2013)
Eagleson M.
Concise Encyclopedia Chemistry, 93-93 (1994)
Eagleson M.
Concise Encyclopedia Chemistry, 124-124 (1994)
Eagleson M.
Concise Encyclopedia Chemistry, 199-199 (1994)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service