Skip to Content
Merck
All Photos(1)

Key Documents

Y0001578

Sildenafil citrate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Sildenafil citrate salt, 1-[[3-(4,7-Dihydro-1-methyl-7-oxo-3-propyl-1H-pyrazolo[4,3-d]pyrimidin-5-yl)-4-ethoxyphenyl]sulfonyl]-4-methylpiperazine citrate salt, 5-[2-Ethoxy-5-(4-methylpiperazin-1-yl)sulfonylphenyl]-1-methyl-3-propyl-4H-pyrazolo[5,4-e]pyrimidin-7-one citrate salt, Revatio, UK-92,480

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H30N6O4S ·C6H8O7
CAS Number:
Molecular Weight:
666.70
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

sildenafil

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

OC(=O)CC(O)(CC(O)=O)C(O)=O.CCCc1nn(C)c2C(=O)NC(=Nc12)c3cc(ccc3OCC)S(=O)(=O)N4CCN(C)CC4

InChI

1S/C22H30N6O4S.C6H8O7/c1-5-7-17-19-20(27(4)25-17)22(29)24-21(23-19)16-14-15(8-9-18(16)32-6-2)33(30,31)28-12-10-26(3)11-13-28;7-3(8)1-6(13,5(11)12)2-4(9)10/h8-9,14H,5-7,10-13H2,1-4H3,(H,23,24,29);13H,1-2H2,(H,7,8)(H,9,10)(H,11,12)

InChI key

DEIYFTQMQPDXOT-UHFFFAOYSA-N

Gene Information

human ... PDE5A(8654)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Sildenafil citrate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Sildenafil is a potent, selective inhibitor of cGMP specific phosphodiesterase type 5 (PDE5). Sildenafil is used to treat erectile dysfunction and pulmonary arterial hypertension. NO activates guanylate cyclase, which results in increased levels of cGMP, producing smooth muscle relaxation. Sildenafil enhances the effect of NO by inhibiting PDE5, which is responsible for degradation of cGMP.
Sildenafil is a potent, selective inhibitor of cGMP specific phosphodiesterase type 5 (PDE5). Sildenafil is used to treat erectile dysfunction and pulmonary hypertension.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

U Ozturk et al.
Irish journal of medical science, 183(3), 449-453 (2013-11-06)
It has been shown on experimental rat models that type 5-phosphodiesterase isoenzyme (PDE5) inhibitors have anti-fibrotic effects for Peyronie's disease (PD); however, this issue has not been addressed clinically. The aim of this study was to document the effects of
M Paranjpe et al.
International journal of pharmaceutics, 476(1-2), 41-49 (2014-09-28)
For the development of any colloidal system, thorough characterization is extremely essential. This article discusses the physicochemical characterization of sildenafil-loaded solid lipid nanoparticle dispersions (SLN) including stability analysis over 6 months time period for possible pulmonary administration for the treatment
Jing Peng et al.
Urology, 84(6), 1389-1394 (2014-10-13)
To evaluate the results of nocturnal penile erection test and response to daily sildenafil in patients with erectile dysfunction (ED) due to pelvic fracture urethral disruption. From January 2010 to January 2012, we included 38 patients with ED due to
Lisa Nivison-Smith et al.
Experimental eye research, 128, 43-56 (2014-09-23)
Sildenafil, the active ingredient in Viagra, has been reported to cause transient visual disturbance from inhibition of phosphodiesterase 6 (PDE6), a key enzyme in the visual phototransduction pathway. This study investigated the effects of sildenafil on the rd1(+/-) mouse, a
M W Helmy et al.
Journal of physiology and pharmacology : an official journal of the Polish Physiological Society, 65(3), 393-399 (2014-06-17)
Although the protective role of either nitric oxide (NO) or endothelin (ET) receptors modulation on the severity of cisplatin-induced nephrotoxicity has been recognized in previous studies including our own, the possible interaction between the two pathways remains obscure. In this

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service