Skip to Content
Merck
All Photos(1)

Key Documents

Y0001197

Cetirizine for peak identification

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Cetirizine dihydrochloride, [2-[4-[(4-Chlorophenyl)phenylmethyl]-1-piperazinyl]ethoxy]acetic acid dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H25ClN2O3 · 2HCl
CAS Number:
Molecular Weight:
461.81
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

cetirizine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

OC(COCCN1CCN(C(C2=CC=C(Cl)C=C2)C3=CC=CC=C3)CC1)=O.[H]Cl.[H]Cl

InChI

1S/C21H25ClN2O3.2ClH/c22-19-8-6-18(7-9-19)21(17-4-2-1-3-5-17)24-12-10-23(11-13-24)14-15-27-16-20(25)26;;/h1-9,21H,10-16H2,(H,25,26);2*1H

InChI key

PGLIUCLTXOYQMV-UHFFFAOYSA-N

Gene Information

human ... HRH1(3269)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Cetirizine for peak identification EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Cetirizine hydrochloride is an orally active and selective H1-receptor antagonist. Antihistaminic; Piperazines. Non-sedating type histamine H1-receptor antagonist; major metabolite of hydroxyzine. Pharmacological activity resides primarily in the (R)-isomer.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Ko-Haung Lue et al.
International journal of pediatric otorhinolaryngology, 76(7), 994-1001 (2012-04-20)
Supplementary consumption of probiotics may temporarily alter the intestinal microflora of infants and children, thereby preventing and treating allergic disorders. To compare the clinical efficacy of levocetirizine with that of levocetirizine plus Lactobacillus johnsonii EM1 (Lj EM1) for treating perennial
Macheng Yan et al.
Journal of traditional Chinese medicine = Chung i tsa chih ying wen pan, 33(1), 60-64 (2013-04-20)
To investigate the potential rules and knowledge of Traditional Chinese Medicine (TCM) and Western Medicine (WM) treatment on chronic urticaria (CU) based on data-mining methods. Sixty patients with chronic urticaria, treated with TCM and WM, were selected. Gray correlation analyses
Arnold Bahlmann et al.
Chemosphere, 89(11), 1278-1286 (2012-06-12)
Carbamazepine (CBZ), caffeine and cetirizine were monitored by enzyme-linked immunosorbent assays (ELISAs) in surface and wastewaters from Berlin, Germany. This fast and cost-efficient method enabled to assess the spatial and temporal variation of these anthropogenic markers in a high-throughput screening.
Susan D Mathias et al.
Allergy and asthma proceedings, 33(2), 186-190 (2012-04-25)
The Urticaria Patient Daily Diary, including the Urticaria Activity Score, has recently been validated in adults with chronic idiopathic urticaria (CIU), but its validity in adolescents is unknown. This study was designed to (1) assess the content validity of the
Natalie J Carter
Drugs, 72(9), 1257-1269 (2012-06-13)
Bilastine is an orally administered, second-generation antihistamine used in the symptomatic treatment of seasonal or perennial allergic rhinoconjunctivitis and urticaria. In two well designed phase III trials, 14 days' treatment with bilastine was associated with a significantly lower area under

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service