Skip to Content
Merck
All Photos(1)

Key Documents

Y0000615

Enalaprilat dihydrate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

(2S)-1-[(2S)-2-[[(1S)-1-Carboxy-3-phenyl-propyl]amino]propanoyl]pyrrolidine-2-carboxylic acid, Vasotec

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H24N2O5 ·2H2O
CAS Number:
Molecular Weight:
384.42
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

enalaprilat

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

O.O.C[C@H](N[C@@H](CCc1ccccc1)C(O)=O)C(=O)N2CCC[C@H]2C(O)=O

InChI

1S/C18H24N2O5.2H2O/c1-12(16(21)20-11-5-8-15(20)18(24)25)19-14(17(22)23)10-9-13-6-3-2-4-7-13;;/h2-4,6-7,12,14-15,19H,5,8-11H2,1H3,(H,22,23)(H,24,25);2*1H2/t12-,14-,15-;;/m0../s1

InChI key

MZYVOFLIPYDBGD-MLZQUWKJSA-N

Gene Information

human ... ACE(1636)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Enalaprilat dihydrate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Enalaprilat is an inhibitor of angiotensin converting enzyme (ACE), antihypertensive, and a Bradykinin B1 receptor activator. Enalaprilat has nM potency versus ACE and also activates B1 receptors to release NO.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enalaprilat.
Dimensions of critical care nursing : DCCN, 19(2), 22-22 (2000-07-06)
K Chatterjee et al.
Drugs, 39 Suppl 4, 29-40 (1990-01-01)
Enalapril is an effective angiotensin-converting enzyme (ACE) inhibitor which produces salutary, beneficial haemodynamic effects in patients with congestive heart failure. Enalapril also produces beneficial neurohumoral changes, as well as improving abnormal coronary haemodynamics and myocardial energetics that are frequently encountered
Kristy L Jackson et al.
Hypertension (Dallas, Tex. : 1979), 63(4), 811-818 (2014-01-22)
BPH/2J mice are recognized as a neurogenic model of hypertension primarily based on overactivity of the sympathetic nervous system and greater neuronal activity in key autonomic cardiovascular regulatory brain regions. The medial amygdala (MeAm) is a forebrain region that integrates
Brian C Ferslew et al.
Drug metabolism and disposition: the biological fate of chemicals, 42(9), 1567-1574 (2014-06-25)
Hepatic uptake and efflux transporters govern the systemic and hepatic exposure of many drugs and metabolites. Enalapril is a pharmacologically inactive prodrug of enalaprilat. Following oral administration, enalapril is converted to enalaprilat in hepatocytes and undergoes translocation into the systemic
Sylwia Magiera et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 980, 79-87 (2015-01-16)
Given the increasing popularity of aliskiren, particularly in combination with angiotensin converting enzyme inhibitor (e.g. enalapril), it is important to determine whether its use in combination with these agents is associated with potentially life threatening safety events. Analytical methods for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service