Skip to Content
Merck
All Photos(1)

Key Documents

S0695000

Sodium amidotrizoate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Sodium diatrizoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H8I3N2NaO4
CAS Number:
Molecular Weight:
635.90
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

sodium amidotrizoate

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[Na+].Ic1c(c(c(c(c1NC(=O)C)I)C(=O)[O-])I)NC(=O)C

InChI

1S/C11H9I3N2O4.Na/c1-3(17)15-9-6(12)5(11(19)20)7(13)10(8(9)14)16-4(2)18;/h1-2H3,(H,15,17)(H,16,18)(H,19,20);/q;+1/p-1

InChI key

ZEYOIOAKZLALAP-UHFFFAOYSA-M

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Sodium amidotrizoate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Karina A Mathes et al.
PloS one, 14(8), e0221050-e0221050 (2019-08-23)
The aim of this study was to describe the specific gross and radiographic anatomy of the digestive tract of inland bearded dragons (Pogona vitticeps). Eleven bearded dragon cadavers of both sexes (6 females, 5 males) were dissected to examine, measure
Serdar Arslan et al.
Japanese journal of radiology, 35(6), 296-302 (2017-03-11)
To demonstrate the successful percutaneous treatment of extrahepatic cystic echinococcosis as an alternative to surgical procedures. A total of 27 extrahepatic hydatid cysts in 12 patients, the spleen in 8 patients, muscles and soft tissues in 3 patients each and
Giancarlo Mansueto et al.
Cardiovascular and interventional radiology, 43(2), 302-310 (2019-11-22)
To investigate the effectiveness of percutaneous approaches to treat bile leak and to propose an anatomical classification of biliary fistula to guide the most appropriate percutaneous approach. Fifty-six patients with bile leakage after hepatobiliary surgery were included. Based on preoperative

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service