Skip to Content
Merck
All Photos(1)

Key Documents

PHL89516

Scopoletin

phyproof® Reference Substance

Synonym(s):

6-Methoxyumbelliferone, 6-Methylesculetin, 7-Hydroxy-6-methoxy-2H-1-benzopyran-2-one, 7-Hydroxy-6-methoxycoumarin, Buxuletin, Chrysatropic acid, Escopoletin, Esculetin-6-methyl ether, Gelseminic acid, Murrayetin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H8O4
CAS Number:
Molecular Weight:
192.17
Beilstein:
156296
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥95.0% (HPLC)

manufacturer/tradename

PhytoLab

mp

203-205 °C (lit.)

application(s)

food and beverages

format

neat

SMILES string

COc1cc2C=CC(=O)Oc2cc1O

InChI

1S/C10H8O4/c1-13-9-4-6-2-3-10(12)14-8(6)5-7(9)11/h2-5,11H,1H3

InChI key

RODXRVNMMDRFIK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Application



  • Thiocyanate-Treated Perovskite-Nanocrystal-Based Light-Emitting Diodes with Insight in Efficiency Roll-Off.: This study explores the applications of ammonium thiocyanate solution in the enhancement of light-emitting diode performance, offering potential insights into improving efficiency and understanding the mechanisms of efficiency roll-off (Chen et al., 2020).


  • Flow-injection simultaneous determination of iron(III) and copper(II) and of iron(III) and palladium(II) based on photochemical reactions of thiocyanato-complexes.: Research demonstrating the use of ammonium thiocyanate in photochemical methods to simultaneously determine metal ions, showcasing a novel application in analytical chemistry (Oguma et al., 2002).


  • The thiocyanate titration of mercury and the standardization of ammonium thiocyanate solution.: This article covers the development of methodologies for the titration of mercury using ammonium thiocyanate, highlighting its significance in quantitative chemical analysis (Cumming et al., 1952).


  • Specific determination of ferbam residues in fog-water.: This research applies ammonium thiocyanate for the specific determination of ferbam residues in environmental samples, enhancing the techniques available for environmental monitoring and safety assessments (Agarwal et al., 2003).




Biochem/physiol Actions

Scopoletin significantly increases activity of lipoprotein lipase in adipocytes by promoting transcription of the LPL gene. It also partially reverses inhibition of LPL by TNF-α.

Other Notes

This compound is commonly found in plants of the genus: achillea angelica glycyrrhiza lycium pimpinella

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service