Skip to Content
Merck
All Photos(3)

Documents

P5654

Sigma-Aldrich

Piroxicam

≥98% (TLC)

Synonym(s):

4-Hydroxy-2-methyl-3-(pyrid-2-yl-carbamoyl)-2H-1,2-benzothiazine 1,1-dioxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H13N3O4S
CAS Number:
Molecular Weight:
331.35
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.77

biological source

synthetic (organic)

Quality Level

Assay

≥98% (TLC)

form

powder

storage temp.

2-8°C

SMILES string

CN1C(C(=O)Nc2ccccn2)=C(O)c3ccccc3S1(=O)=O

InChI

1S/C15H13N3O4S/c1-18-13(15(20)17-12-8-4-5-9-16-12)14(19)10-6-2-3-7-11(10)23(18,21)22/h2-9,19H,1H3,(H,16,17,20)

InChI key

QYSPLQLAKJAUJT-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

Piroxicam has been used:
  • for the induction of colitis in mice
  • as a non-steroidal anti-inflammatory drug to increase the sensitivity to gut injury in interleukin-10 (IL-10) knockout mice
  • as a chemopreventive agent and as a positive control to study its effects on tumor growth in mice small intestine

Biochem/physiol Actions

Piroxicam is a non‐steroidal anti‐inflammatory drug (NSAID). It possesses analgesic properties and exhibits therapeutic effects against rheumatic disorders.
Cyclooxygenase inhibitor.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - STOT RE 2 Oral

Target Organs

Gastrointestinal tract

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T J Carty et al.
Prostaglandins, 19(1), 51-59 (1980-01-01)
The new non-steroidal antiinflammatory (NSAI)2 agent, piroxicam [4-hydroxy-2-methyl-N-(2-pyridyl)-2H-1,2-benzothiazine-3-carboxamide 1,1-dioxide], is a highly active inhibitor of prostaglandin (PG) synthesis by methylcholanthrene transformed mouse fibroblasts (MC5-5) and rabbit synovial cells in culture. Comparison of the PG biosynthesis inhibitory activity of piroxicam with
Thymoquinone attenuates tumor growth in ApcMin mice by interference with Wnt-signaling
Lang M, et al.
Molecular Cancer, 12(1), 41-41 (2013)
Michaela Lang et al.
Molecular cancer, 12(1), 41-41 (2013-05-15)
Patients with familial adenomatous polyposis (FAP) are at increased risk for the development of colorectal cancer. Surgery and chemoprevention are the most effective means to prevent cancer development. Thymoquinone (TQ) is considered the main compound of the volatile Nigella sativa
Jiawei Guo et al.
Cell reports. Medicine, 4(1), 100881-100881 (2023-01-06)
Systematic bone loss is commonly complicated with inflammatory bowel diseases (IBDs) with unclear pathogenesis and uncertain treatment. In experimental colitis mouse models established by dextran sulfate sodium and IL-10 knockout induced with piroxicam, bone mass and quality are significantly decreased.
Seung-Won Choi et al.
Plant biotechnology (Tokyo, Japan), 39(3), 323-327 (2022-11-10)
Agrobacterium-mediated transformation is a key innovation for plant breeding, and routinely used in basic researches and applied biology. However, the transformation efficiency is often the limiting factor of this technique. In this study, we discovered that oxicam-type nonsteroidal anti-inflammatory drugs

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service