Skip to Content
Merck
All Photos(1)

Key Documents

D1750000

Diflunisal

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

2′,4′-Difluoro-4-hydroxy-[1,1′-biphenyl]-3-carboxylic acid, 5-(2,4-Difluorophenyl)salicylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H8F2O3
CAS Number:
Molecular Weight:
250.20
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

diflunisal

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

FC1=CC(F)=CC=C1C2=CC(C(O)=O)=C(O)C=C2

InChI

1S/C13H8F2O3/c14-8-2-3-9(11(15)6-8)7-1-4-12(16)10(5-7)13(17)18/h1-6,16H,(H,17,18)

InChI key

HUPFGZXOMWLGNK-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Diflunisal EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M W Rich et al.
Chest, 111(6), 1767-1769 (1997-06-01)
Drug-induced pneumonitis is an uncommon complication of nonsteroidal anti-inflammatory drug administration. Herein is the first reported case of pneumonitis resulting from diflunisal therapy. The patient demonstrated clinical and biopsy evidence of systemic vasculitis. She responded dramatically to administration of systemic
R N Brogden et al.
Drugs, 19(2), 84-106 (1980-02-01)
Diflunisal is a salicylic acid derivative with analgesic and anti-inflammatory activity. It has been studied in osteoarthritis, pain resulting from musculoskeketal sprains and strains and from minor surgery and cancer. The duration of its analgesic effect is longer than that
Jack O Wasey et al.
The Cochrane database of systematic reviews, (4)(4), CD007440-CD007440 (2010-04-16)
Diflunisal is a long-acting non-steroidal anti-inflammatory drug (NSAID) most commonly used to treat acute postoperative pain or chronic joint pain from osteoarthritis and rheumatoid arthritis. This review analyses the effectiveness and harm of different doses of diflunisal in the context
G M Lawton et al.
Australian dental journal, 38(4), 265-271 (1993-08-01)
Diflunisal is frequently used for management of postoperative dental pain as it has some advantages over other commonly used non-steroidal anti-inflammatory drugs including improved analgesia, a prolonged duration of action lasting up to 12 hours, and a low incidence of
R O Davies
Pharmacotherapy, 3(2 Pt 2), 9S-22S (1983-03-01)
Diflunisal is a new salicylic acid derivative identified after a search for a compound with improved potency, enhanced gastrointestinal tolerance relative to its antiinflammatory activity, and a longer duration of action than that of aspirin. Animal and clinical studies have

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service