Skip to Content
Merck
All Photos(1)

Key Documents

C2385000

Clonazepam

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

5-(2-Chlorophenyl)-7-nitro-3H-1,4-benzodiazepin-2(1H)-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H10ClN3O3
CAS Number:
Molecular Weight:
315.71
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

clonazepam

manufacturer/tradename

EDQM

drug control

USDEA Schedule IV; regulated under CDSA - not available from Sigma-Aldrich Canada; psicótropo (Spain); Decreto Lei 15/93: Tabela IV (Portugal)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Clc1ccccc1C2=NCC(=O)Nc3ccc(cc23)N(=O)=O

InChI

1S/C15H10ClN3O3/c16-12-4-2-1-3-10(12)15-11-7-9(19(21)22)5-6-13(11)18-14(20)8-17-15/h1-7H,8H2,(H,18,20)

InChI key

DGBIGWXXNGSACT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Clonazepam EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shigeru Morishita
Human psychopharmacology, 24(3), 191-198 (2009-03-31)
Clonazepam, first used for seizure disorders, is now increasingly used to treat affective disorders. We summarize the use of clonazepam to improve the management of depression. Clonazepam is useful for treatment-resistant and/or protracted depression, as well as for acceleration of
Stuart J McCarter et al.
Sleep medicine, 14(3), 237-242 (2013-01-29)
REM sleep behavior disorder (RBD) is usually characterized by potentially injurious dream enactment behaviors (DEB). RBD treatment aims to reduce DEBs and prevent injury, but outcomes require further elucidation. We surveyed RBD patients to describe longitudinal treatment outcomes with melatonin
Jerrold F Rosenbaum
The Journal of clinical psychiatry, 65 Suppl 5, 3-6 (2004-04-14)
The pathophysiology of anxiety disorders is not clearly understood; therefore, clinical observation, case reports, and case reviews continue to enhance physicians' understanding of disease and treatment mechanisms. At Massachusetts General Hospital (MGH), physicians and researchers are guided by the recognition
Raffaele Ferri et al.
Sleep medicine, 14(1), 24-29 (2012-10-27)
To analyze the differences in sleep structure and nocturnal motor activity between drug-free REM sleep behavior disorder (RBD) patients and those under therapy with clonazepam, and to evaluate the long-term longitudinal changes under continued therapy with clonazepam. Fifty-seven consecutive iRBD
G Chouinard
The Journal of clinical psychiatry, 48 Suppl, 29-37 (1987-10-01)
This is a report on the efficacy of clonazepam in the treatment of acute mania. The advantages of clonazepam over standard neuroleptics are its rapidity of action, its lack of toxicity, and, particularly important, the fact that it does not

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service