Skip to Content
Merck
All Photos(3)

Documents

89700

Supelco

Toluene-3,4-dithiol

for spectrophotometric det. of Mo, Sn, W, and also Ag and Re, ≥97.0%

Synonym(s):

3,4-Dimercaptotoluene, 4-Methyl-1,2-benzenedithiol, ‘Dithiol’

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H3(SH)2
CAS Number:
Molecular Weight:
156.27
Beilstein:
1635270
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NA.21

Quality Level

Assay

≥97.0% (GC)
≥97.0%

form

powder

quality

for spectrophotometric det. of Mo, Sn, W, and also Ag and Re

technique(s)

UV/Vis spectroscopy: suitable

bp

135-137 °C/17 mmHg (lit.)

mp

28-32 °C
30-33 °C (lit.)

density

1.179 g/mL at 25 °C (lit.)

storage temp.

−20°C

SMILES string

Cc1ccc(S)c(S)c1

InChI

1S/C7H8S2/c1-5-2-3-6(8)7(9)4-5/h2-4,8-9H,1H3

InChI key

NIAAGQAEVGMHPM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Toluene-3,4-dithiol with molybdenum(VI) in mineral acid forms dark-green coloured complex. It is purified by vacuum distillation under nitrogen.

Application

Toluene-3,4-dithiol may be suitable as colorimetric reagent for the determination of technetium using spectrophotometry.

Other Notes

Reagent for the spectrophotometric determination of Mo, Sn, W, and also Ag and Re

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K.L. Cheng et al. et al.
Handbook of Organic Analytical Reagents null
Kuang Lu Cheng, Keihei Ueno, Toshiaki Imamur
CRC Handbook of Organic Analytical Reagents, 474-474 (1992)
W John Williams
Handbook of Anion Determination, 113-113 (2013)
E P Welsch
Talanta, 30(11), 876-878 (1983-11-01)
1-g sample is decomposed with nitric and hydrofluoric acids, and after evaporation of the solution to dryness the residue is dissolved in concentrated hydrochloric acid. A clear aliquot is treated with stannous chloride to inhibit interferences. The blue tungsten dithiol
Spectrophotometric Determination of Technetium with Toluene-3, 4-dithiol.
Miller, FRANCIS J., and PAUL F. Thomason.
Analytical Chemistry, 33, 404-406 (1961)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service