Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
100 tests | Check Cart for Availability | $1,950.00 |
About This Item
pathogen testing
Product Name
GDS E. coli O157:H7 Tq, BioControl, Molecular based PCR test for the detection of E. coli O157:H7 in food and environmental samples
Quality Segment
agency
Immunomagnetic Separation (IMS), AOAC® (OMA), AOAC® (PTM), Health Canada, MicroVal
product line
Assurance GDS®
shelf life
6‑12 mo.
packaging
pkg of 100
manufacturer/tradename
BioControl Systems 71007-100
technique(s)
PCR: suitable
application(s)
food and beverages
pathogen testing
compatibility
BioControl GDS, for use with GDS Rotor-Gene
storage temp.
2-8°C
suitability
Escherichia coli (O157:H7)
General description
Application
Features and Benefits
- Results available as fast as 8 hours for 25 g samples / 10 hours for composite 375 g samples.
- Positive samples can be confirmed for stx genes in an additional 2 hours with GDS Shiga Toxin Genes (O157) assay (PN 71005BC).
- Validated for ground beef, beef trim, leafy greens, juices, and sprout irrigation water.
- Features innovative PickPen® immunomagnetic separation (IMS) for additional specificity and fast time to results.
- Simplified sample prep does not require heating blocks or preparation of reagents.
Other Notes
- E. coli O157:H7 Amplification Tubes Tq (0.2 mL)
- O157 Concentration Reagent
- Resuspension Buffer Tq
- Wash Solution
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| manufacturer/tradename BioControl Systems 71007-100 | manufacturer/tradename BioControl Systems 71007-576ATM | manufacturer/tradename BioControl Systems | manufacturer/tradename BioControl Systems 71010-100 |
| suitability Escherichia coli (O157:H7) | suitability Escherichia coli (O157:H7) | suitability Escherichia coli (O157:H7) | suitability Listeria monocytogenes |
| compatibility BioControl GDS, for use with GDS Rotor-Gene | compatibility BioControl GDS, for use with GDS Rotor-Gene | compatibility BioControl GDS, for use with GDS Rotor-Gene | compatibility BioControl GDS, for use with GDS Rotor-Gene |
| Quality Level 200 | Quality Level 200 | Quality Level 200 | Quality Level 200 |
| agency Immunomagnetic Separation (IMS), AOAC® (PTM), MicroVal, AOAC® (OMA), Health Canada | agency PCR, Immunomagnetic Separation (IMS), AOAC® (OMA), AOAC® (PTM), Health Canada, MicroVal | agency PCR, Immunomagnetic Separation (IMS), according to AOAC® (PTM) | agency PCR, Immunomagnetic Separation (IMS), AOAC® (PTM), Health Canada, MicroVal |
| application(s) food and beverages | application(s) food and beverages | application(s) food and beverages | application(s) food and beverages |
Storage Class
10 - Combustible liquids
signalword
Danger
hcodes
Hazard Classifications
ED ENV 1
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



