Skip to Content
Merck
All Photos(1)

Documents

33630

Sigma-Aldrich

Dibenzyl ether

purum, ≥98.0% (GC)

Synonym(s):

Benzyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5CH2)2O
CAS Number:
Molecular Weight:
198.26
Beilstein:
1911156
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

purum

Quality Level

Assay

≥98.0% (GC)

form

liquid

refractive index

n20/D 1.562

bp

158-160 °C/10 mmHg (lit.)

mp

1.5-3.5 °C (lit.)

density

1.043 g/mL at 20 °C (lit.)

SMILES string

C(OCc1ccccc1)c2ccccc2

InChI

1S/C14H14O/c1-3-7-13(8-4-1)11-15-12-14-9-5-2-6-10-14/h1-10H,11-12H2

InChI key

MHDVGSVTJDSBDK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Thermolysis of dibenzyl ether in water was studied to probe into volatiles obtained from the reaction of coal in supercritical water. Diffusion coefficients of dibenzyl ether at infinite dilution in CO2 and ethanol mixtures has been measured.

Application

  • Green synthesis route: Research demonstrated a green synthesis route using dibenzyl ether for the formation of benzyl phenyl sulfide, emphasizing environmentally friendly methods in chemical synthesis processes (Wu et al., 2023).
  • Nanoparticle synthesis by thermal decomposition: Dibenzyl ether was used in the thermal decomposition process for the synthesis of manganese oxide nanoparticles, showcasing its utility in facilitating controlled chemical reactions (Martinez de la Torre and Bennewitz, 2020).

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1B

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

278.6 °F - Pensky-Martens closed cup

Flash Point(C)

137 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dibenzyl ether as a probe into the supercritical fluid solvent extraction of volatiles from coal with water.
Townsend SH and Klein MT.
Fuel: The Science and Technology of Fuel and Energy, 64(5), 635-638 (1985)
Diffusion of benzyl acetate, 2-phenylethyl acetate, 3-phenylpropyl acetate, and dibenzyl ether in mixtures of carbon dioxide and ethanol.
Suarez-Iglesias O, et al.
Industrial & Engineering Chemistry Research, 46(11), 3810-3819 (2007)
Kulandaivelu Sivanandan et al.
Journal of the American Chemical Society, 127(7), 2020-2021 (2005-02-17)
Dendrons with a fluorescent probe in a specific location have been synthesized and characterized. Accessibility of guest molecules to each of these layers was then analyzed using an intermolecular photoinduced electron-transfer process. Comparisons of the accessibility among each layer of
Klaus Becker et al.
PloS one, 7(3), e33916-e33916 (2012-04-06)
Generally, chemical tissue clearing is performed by a solution consisting of two parts benzyl benzoate and one part benzyl alcohol. However, prolonged exposure to this mixture markedly reduces the fluorescence of GFP expressing specimens, so that one has to compromise
G A Burdock et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 30(7), 559-566 (1992-07-01)
Dibenzyl ether (FEMA No. 2371, CAS No. 103-50-4) was given in the diet to rats at a rate of 62, 196 or 620 mg/kg/day for 91 consecutive days. Body weights and food consumption were measured weekly; haematological, clinical chemistry and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service