Skip to Content
Merck
All Photos(3)

Documents

181838

Sigma-Aldrich

Silicone oil

viscosity 60,000 cSt (25 °C)

Synonym(s):

Dimethyl polysiloxane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[-Si(CH3)2O-]n
CAS Number:
MDL number:
UNSPSC Code:
12352100
NACRES:
NA.21

vapor density

>1 (vs air)

Quality Level

vapor pressure

<5 mmHg ( 25 °C)
5 mmHg ( 20 °C)

manufacturer/tradename

(Xiameter® PMX-200 fluid)

refractive index

n20/D 1.403 (lit.)

viscosity

60,000 cSt(25 °C)

bp

>140 °C/0.002 mmHg (lit.)

density

0.98 g/mL at 25 °C

Looking for similar products? Visit Product Comparison Guide

General description

Xiameter PMX-200

Application

  • Enhanced performance coatings: A study utilized silicone oil to develop regenerative superhydrophobic coatings, aimed at improving the performance and durability of high-voltage electrical insulators in cold climates. This application highlights the material′s effectiveness in environmental protection and infrastructure longevity (Khademsameni et al., 2024).
  • Durable super-hydrophobicity: Silicone oil was grafted onto rough thermoplastic polyurethane surfaces to create durable super-hydrophobic properties, demonstrating a significant advancement in materials engineering for creating water-resistant surfaces (Tan et al., 2024).

Legal Information

XIAMETER is a registered trademark of The Dow Chemical Company or an affiliated company of Dow

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

214.0 °F - closed cup

Flash Point(C)

101.1 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Karthik R Balakrishnan et al.
Lab on a chip, 13(7), 1302-1307 (2013-02-07)
Resistive-pulse sensing (RPS), which is based on measuring the current pulse produced when a single particle transits a pore or channel, is an extremely versatile technique used to determine the size and concentration of cells and viruses and to detect
John R Fowler et al.
The Orthopedic clinics of North America, 44(3), 425-431 (2013-07-06)
Radial head fractures without associated bony or ligamentous injury can be safely treated with internal fixation, if possible, or arthroplasty if nonreconstructable. However, nonreconstructable radial head fractures in association with elbow dislocation and/or ligamentous injury in the elbow or forearm
Kristina Kreppenhofer et al.
Langmuir : the ACS journal of surfaces and colloids, 29(11), 3797-3804 (2013-02-23)
Here we demonstrate the generation of polymer monolithic surfaces possessing a gradient of pore and polymer globule sizes from ~0.1 to ~0.5 μm defined by the composition of two polymerization mixtures injected into a microfluidic chip. To generate the gradient
M Krutyeva et al.
Physical review letters, 110(10), 108303-108303 (2013-03-26)
We present neutron spin echo experiments that address the much debated topic of dynamic phenomena in polymer melts that are induced by interacting with a confining surface. We find an anchored surface layer that internally is highly mobile and not
Nobuo Yoshimoto et al.
Scientific reports, 3, 1191-1191 (2013-02-05)
When establishing the most appropriate cells from the huge numbers of a cell library for practical use of cells in regenerative medicine and production of various biopharmaceuticals, cell heterogeneity often found in an isogenic cell population limits the refinement of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service