Skip to Content
Merck
All Photos(1)

Key Documents

14631

Supelco

(±)-Jasmonic acid

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H18O3
CAS Number:
Molecular Weight:
210.27
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

Assay

≥90.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

CC\C=C/C[C@H]1[C@@H](CCC1=O)CC(O)=O

InChI

1S/C12H18O3/c1-2-3-4-5-10-9(8-12(14)15)6-7-11(10)13/h3-4,9-10H,2,5-8H2,1H3,(H,14,15)/b4-3-/t9-,10-/m0/s1

InChI key

ZNJFBWYDHIGLCU-CMIOBCHKSA-N

Looking for similar products? Visit Product Comparison Guide

General description

(±)-Jasmonic acid is a plant lipid derivative[1] and a naturally available growth regulator found in higher plants.[2]

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qingyuan Li et al.
Scientific reports, 6, 36463-36463 (2016-11-08)
Camellia is a well-known ornamental flower native to Southeast of Asia, including regions such as Japan, Korea and South China. However, most species in the genus Camellia are cold sensitive. To elucidate the cold stress responses in camellia plants, we
Jasmonic acid distribution and action in plants: regulation during development and response to biotic and abiotic stress.
Creelman AR and Mullet EJ
Proceedings of the National Academy of Sciences of the USA, 92(10), 4114-4119 (1995)
R A Creelman et al.
Proceedings of the National Academy of Sciences of the United States of America, 89(11), 4938-4941 (1992-06-01)
Jasmonic acid (JA) and its methyl ester, methyl jasmonate (MeJA), are plant lipid derivatives that resemble mammalian eicosanoids in structure and biosynthesis. These compounds are proposed to play a role in plant wound and pathogen responses. Here we report the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service