Skip to Content
Merck
All Photos(1)

Key Documents

DIUTP-RO

Roche

Digoxigenin-11-dUTP, alkali-labile

=85% (HPLC), solution, pkg of 25 μL (11573152910 [1 mM]), pkg of 125 μL (11573179910 [1 mM]]])

Synonym(s):

Digoxigenin-11-dUTP, alkali-labile, digoxigenin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105500

Quality Level

Assay

85% (HPLC)

form

solution

packaging

pkg of 125 μL (11573179910 [1 mM]]])
pkg of 25 μL (11573152910 [1 mM])

manufacturer/tradename

Roche

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Digoxigenin (DIG)-11-deoxyuridine triphosphate (dUTP), alkali-labile is provided as in 1 mM tetralithium salt, solution. In which DIG is bound to dUTP via an alkali-sensitive ester linkage.

Application

Digoxigenin-11-dUTP, alkali-labile can be used for non-radioactive DNA labeling e.g. random primed or nick translation. It can also be used for labeling probes which are preferred in hybridization experiments where stripping and reprobing of the membrane is intended.

Biochem/physiol Actions

Digoxigenin (DIG)-11-deoxyuridine triphosphate (dUTP) replaces deoxythymidine triphophate (dTTP) in the random-primed DNA labeling reaction or in nick translation in a ratio of 35% DIG-11-dUTP and 65% dTTP. It is ideal as a substrate for DNA polymerase, Taq DNA polymerase, terminal transferase and reverse transcriptase.

Other Notes

For life science research only. Not for use in diagnostic procedures.
When this nucleotide is subjected to 0.1 - 0.5 NaOH at +15 to +25°C, the DIG moiety will be removed. It is therefore ideal for use in DIG-labeled probes that can be removed by alkali denaturation (e.g ., in stripping and reprobing of blots). However, if you intend to make a DIG-labeled DNA that will survive alkaline treatment, do not use this alkali-labile preparation. Instead, use the alkali-stable form of DIG-11-dUTP.

also commonly purchased with this product

Product No.
Description
Pricing

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Araceli Nora García et al.
BMC plant biology, 14, 248-248 (2014-09-18)
The production of antimicrobial peptides is a common defense strategy of living cells against a wide range of pathogens. Plant snakin peptides inhibit bacterial and fungal growth at extremely low concentrations. However, little is known of their molecular and ecological
Interstitial telomeric sequence blocks in constitutive pericentromeric heterochromatin from Pyrgomorpha conica (Orthoptera) are enriched in constitutive alkali-labile sites
Lopez-Fernandez, Carmen, et al
Mutation Research. Fundamental and Molecular Mechanisms of Mutagenesis, 599 (1), 36-44 (2006)
James E Dombrowski et al.
BMC research notes, 4, 46-46 (2011-03-08)
Choke, caused by the endophytic fungus Epichloë typhina, is an important disease affecting orchardgrass (Dactylis glomerata L.) seed production in the Willamette Valley. Little is known concerning the conditions necessary for successful infection of orchardgrass by E. typhina. Detection of
Sara Bembich et al.
Nucleic acids research, 42(5), 3362-3371 (2013-12-27)
TDP-43 is a nuclear protein involved in many aspects of RNA metabolism. To ensure cellular viability, its expression levels within cells must be tightly regulated. We have previously demonstrated that TDP-43 autoregulation occurs through the activation of a normally silent
Xingqi Chen et al.
Epigenetics, 9(11), 1439-1445 (2014-12-09)
Despite considerable efforts, our understanding of the organization of higher order chromatin conformations in single cells and how these relate to chromatin marks remains poor. We have earlier invented the Chromatin In Situ Proximity (ChrISP) technique to determine proximities between

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service