Skip to Content
Merck
All Photos(1)

Key Documents

V0CPAK0A1

Milli-Q

VOC-Pak® Polisher

Production of volatile organic compound-free water at the point of dispense of the Milli-Q® IQ/EQ 7 series water purification systems.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104212
eCl@ss:
33050190
NACRES:
JA.13

material

carbon cartridge (activated)

Quality Level

packaging

pkg of 1 unit

manufacturer/tradename

VOC-Pak®

compatibility

for use with Milli-Q® EQ 7000
for use with Milli-Q® EQ 7008
for use with Milli-Q® EQ 7016
for use with Milli-Q® IQ 7000
for use with Milli-Q® IQ 7003
for use with Milli-Q® IQ 7005
for use with Milli-Q® IQ 7010
for use with Milli-Q® IQ 7015

shipped in

ambient

Looking for similar products? Visit Product Comparison Guide

General description

VOC-Pak® Polisher is designed to purify ultrapure water for the analysis of volatile organic compounds (VOCs) in GC and GC-MS analyses. The polisher is made up of activated carbon specifically designed for the effective removal of VOCs from ultrapure water. This unit is an excellent choice for scientists in need of VOC-free ultrapure water for their analytical applications.
VOC-Pak® Polisher produces VOC-free water when fed with ultrapure water at the Q-POD® point of dispense of Milli-Q® IQ 7000, Milli-Q® IQ 7003/05/10/15 and Milli-Q® EQ 7000/08/16 water purification systems.

Application

VOC-Pak® Polisher is used to produce VOC-free water for GC and GC-MS analyses when fed with ultrapure water. The VOC-free ultrapure water delivered is suitable for preparation of blanks, standard solutions, reagents and samples used in the analysis of those substances, or in situations where they may impact results, such as in electrochemistry studies.

Features and Benefits

  • Validated to produce 300 L of VOC-free ultrapure water.
  • Carefree maintenance: the polisher is easily installed and replaced
  • Redesigned bottom tip for a secured installation of the protection bell.
  • e-Sure tag for RFID connection with Q-POD® dispenser enables full traceability (data management) and automatic consumable status monitoring on the POD′s touchscreen interface.
  • Certificate of Quality available online.

Legal Information

Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany
Q-POD is a registered trademark of Merck KGaA, Darmstadt, Germany
VOC-PAK is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ran Chen et al.
Journal of the Electrochemical Society, 163(4), H3032-H3037 (2016-09-20)
Nanoscale scanning electrochemical microscopy (SECM) has emerged as a powerful electrochemical method that enables the study of interfacial reactions with unprecedentedly high spatial and kinetic resolution. In this work, we develop carbon nanoprobes with high electrochemical reactivity and well-controlled size
Tong Sun et al.
Faraday discussions, 210(0), 173-188 (2018-07-07)
This paper is concerned with long-distance interactions between an unbiased metal nanoparticle (NP) and a nanoelectrode employed as a tip in the scanning electrochemical microscope (SECM). A NP immobilized on the inert substrate acts as a bipolar electrode, producing positive
J S Cooper et al.
The Analyst, 140(9), 3233-3238 (2015-03-15)
Partially selective gold nanoparticle sensors have the sensitivity and selectivity to discriminate and quantify benzene, toluene, ethylbenzene, p-xylene and naphthalene (BTEXN) at concentrations relevant to the US Environmental Protection Agency. In this paper we demonstrate that gold nanoparticle chemiresistors can

Related Content

Milli-Q® ultrapure water systems ensure reliable science, with an expanded system offer catering to diverse lab needs and constraints.

Milli-Q® ultrapure water systems ensure reliable science, with an expanded system offer catering to diverse lab needs and constraints.

Milli-Q® ultrapure water systems ensure reliable science, with an expanded system offer catering to diverse lab needs and constraints.

Milli-Q® ultrapure water systems ensure reliable science, with an expanded system offer catering to diverse lab needs and constraints.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service