Skip to Content
Merck
All Photos(2)

Key Documents

SCC134

Sigma-Aldrich

IMG Mouse Microglial Cell Line

Mouse

Synonym(s):

Immortalized microglial cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
eCl@ss:
32011203
NACRES:
NA.81

product name

IMG Mouse Microglial Cell Line, IMG microglial cells recapitulate key features of microglial cell activation and may be used to study neuroinflammatory responses underlying Alzheimer′s disease.

biological source

mouse

Quality Level

technique(s)

cell based assay: suitable

shipped in

ambient

General description

Immortalized MicroGlial Cell Line or IMG is an immortalized microglial cell line derived from adult murine brain infected with the v-raf/v-myc retrovirus. IMG cells express markers that are specific to adult primary microglial cells (F4/80 and CD11b) as assessed by flow cytometry and are responsive to pro- and anti-inflammatory signals (1). They have been shown to phagocytose foreign particles and misfolded proteins such as the amyloid-beta peptide (1). IMG cells are rapidly dividing cells that are typically small and round with few or no processes when cultured in medium containing serum.

Reference:
1. McCarthy RC, Lu DY, Alkhateeb A, Gardeck AM, Lee CH and Wessling-Resnick M (2016). Characterization of a novel adult murine immortalized microglial cell line and its activation by amyloid-beta. J. Neuroinflammation 13: 21
Microglia are the primary immune cells of the central nervous system (CNS), and account for 10-15% of all cells found within the brain. Microglial cells play an important role in maintaining the health of the CNS by the removal of pathogens, infectious agents and damaged cells through phagocytosis. Microglial cells have also been extensively studied for their harmful roles in a variety of neurodegenerative disease including Alzheimer’s disease, Parkinson’s disease and amyotrophic lateral sclerosis.

Cell Line Description

Macrophage Cells

Application

Research Category
Immune Response

Inflammation & Immunology

Neuroscience
Research Sub Category
Neurodegenerative Diseases

Neuroinflammation & Pain
This product is sold solely for research use per the terms of the “Restricted Use Agreement” which govern its use as detailed in the product Data Sheet or Certificate of Analysis. For information regarding any other use, please contact licensing@emdmillipore.com.

Quality

• Each vial contains ≥ 1X106 viable cells.
• Cells are tested negative for infectious diseases by a Mouse Essential CLEAR panel by Charles River Animal Diagnostic Services.
• Cells are verified to be of mouse origin and negative for inter-species contamination from rat, chinese hamster, Golden Syrian hamster, human and non-human primate (NHP) as assessed by a Contamination Clear panel by Charles River Animal Diagnostic Services.
• Cells are negative for mycoplasma contamination.

Storage and Stability

Store in liquid nitrogen. The cells can be cultured for at least 10 passages after initial thawing without significantly affecting the cell marker expression and functionality.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bing-Wen Lu et al.
Advances in ophthalmology practice and research, 3(2), 47-54 (2023-10-17)
Artesunate (ART), a member of the artemisinin family, possesses multi-properties, including anti-inflammation, anti-oxidation, and anti-tumor. ART was recently reported to show anti-neovascularization effect on the cornea, iris, and retina. Compared to the expensive anti-VEGF treatment, this versatile, economical treatment option
Yoshinobu Hirano et al.
Cells, 12(16) (2023-08-26)
We previously demonstrated that neural stem/progenitor cells (NSPCs) were induced within and around the ischemic areas in a mouse model of ischemic stroke. These injury/ischemia-induced NSPCs (iNSPCs) differentiated to electrophysiologically functional neurons in vitro, indicating the presence of a self-repair
Md Rezaul Islam et al.
EMBO molecular medicine, 13(11), e13659-e13659 (2021-10-12)
While some individuals age without pathological memory impairments, others develop age-associated cognitive diseases. Since changes in cognitive function develop slowly over time in these patients, they are often diagnosed at an advanced stage of molecular pathology, a time point when
Akiko Nakano-Doi et al.
International journal of molecular sciences, 24(2) (2023-01-22)
Microglial cells (MGs), originally derived from progenitor cells in a yolk sac during early development, are glial cells located in a physiological and pathological brain. Since the brain contains various cell types, MGs could frequently interact with different cells, such
Zhong-Qing Sun et al.
Neural regeneration research, 17(1), 203-209 (2021-06-09)
Lycium barbarum (LB) is a traditional Chinese medicine that has been demonstrated to exhibit a wide variety of biological functions, such as antioxidation, neuroprotection, and immune modulation. One of the main mechanisms of Alzheimer's disease is that microglia activated by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service