Skip to Content
Merck
All Photos(1)

Key Documents

MABS36

Sigma-Aldrich

Anti-ODC1 Antibody, clone CPTC-ODC1-2

clone CPTC-ODC1-2, from mouse

Synonym(s):

ornithine decarboxylase 1, ornithine decarboxylase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

CPTC-ODC1-2, monoclonal

species reactivity

human

technique(s)

western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ODC1(4953)

General description

Ornithine decarboxylase (ODC1) is the first enzyme in polyamine synthesis and is mandatory for normal growth and is increased in many types of cancers. ODC1 has been shown to suppress the fmc1 deletion in yeast and restores healthy respiratory growth. ODC1 may play a role in promoting DNA synthesis in hepatocytes.

Immunogen

Full length of recombinant protein corresponding to human ODC1.

Material obtained from the National Cancer Institute’s Clinical Proteomic Technologies for Cancer (CPTC) program.

Application

Research Category
Signaling
This Anti-ODC1 Antibody, clone CPTC-ODC1-2 is validated for use in WB for the detection of ODC1.

Quality

Evaluated by Western Blot in MCF-7 cell lysate.

Western Blot Analysis: 10 µg/mL of this antibody detected ODC1 on 10 µg of MCF-7 cell lysate.

Target description

~51 kDa observed. A band at ~100 kDa may be observed in some lysates and may be the homodimer of this protein.

Physical form

Format: Purified
Protein G
Purified mouse monoclonal IgG2aκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
MCF-7 cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Renee C Geck et al.
The Journal of biological chemistry, 295(19), 6263-6277 (2020-03-07)
Treatment of patients with triple-negative breast cancer (TNBC) is limited by a lack of effective molecular therapies targeting this disease. Recent studies have identified metabolic alterations in cancer cells that can be targeted to improve responses to standard-of-care chemotherapy regimens.
Guillaume N Fiches et al.
PLoS pathogens, 18(4), e1010503-e1010503 (2022-04-30)
Polyamines are critical metabolites involved in various cellular processes and often dysregulated in cancers. Kaposi's sarcoma-associated Herpesvirus (KSHV), a defined human oncogenic virus, leads to profound alterations of host metabolic landscape to favor development of KSHV-associated malignancies. In our studies
Milind D Chalishazar et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 25(16), 5107-5121 (2019-06-06)
Small-cell lung cancer (SCLC) has been treated clinically as a homogeneous disease, but recent discoveries suggest that SCLC is heterogeneous. Whether metabolic differences exist among SCLC subtypes is largely unexplored. In this study, we aimed to determine whether metabolic vulnerabilities

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service