Skip to Content
Merck
All Photos(2)

Documents

MAB5308

Sigma-Aldrich

Anti-BACE Antibody, CT, clone 61-3E7

clone 61-3E7, Chemicon®, from mouse

Synonym(s):

ASP2, BACE1, Beta Secretase, beta-Site APP Cleaving Enzyme

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

61-3E7, monoclonal

species reactivity

primate, mouse, human, rat

manufacturer/tradename

Chemicon®

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... BACE1(23621)

General description

Alzheimer′s disease (AD) is characterized by the progressive formation in the brain of insoluble amyloid plaques and vascular deposits consisting of the 4-kD amyloid b-peptide (Ab). Ab generation is initiated by proteolytic cleavage of the amyloid precursor protein (APP) at the N-terminal of Ab by b-secretase. The Ab peptide is then released by proteolytic cleavage at its C-terminus by g-secretase. Because both these proteases are prime candidates for therapeutic intervention, an intense search has been underway to identify these two enzymes.A human transmembrane aspartic-protease (Asp2), referred to as BACE, has been characterized and shown to have all the properties of b-secretase. Four groups in all have now confirmed that BACE (or Asp2) is a convincing candidate for b-secretase.



BACE is an N-glycosylated integral membrane aspartyl protease with Mr=70 kDa. Mature BACE is produced from the immature form through a series of post-translational proteolytic cleavages and glycosylation. Sequence analysis has revealed that the immature form of BACE contains an N-terminal signal sequence (residues 1-21) followed by a large catalytic domain, a single transmembrane domain (residues 461-477), and a short cytoplasmic domain (residues 478-501). The signal sequence (1-21) is cleaved from the immature form by a signal peptidase located in the endoplasmic reticulum (ER), yielding the proBACE protein (Mr=75 kDa) which starts at residue 22. The proBACE protein is modified by cleavage of 24 N-terminal residues (aa 22-45), producing the mature BACE protein.

Specificity

Reacts with BACE (beta-site APP Cleaving Enzyme). Shows no reactivity to BACE2 by Western blot.

Immunogen

Epitope: C-terminus
Synthetic peptide corresponding to the C-terminus of human BACE.

Application

Detect BACE using this Anti-BACE Antibody, C-terminus, clone 61-3E7 validated for use in IP & WB.
Research Category
Neuroscience
Research Sub Category
Neurodegenerative Diseases
Western blotting: 1 μg/mL; recognizes pro and mature forms: ~60-75kDa on reducing westerns. BACE is N-terminally glycosylated which causes the wide size range.

Immunohistochemistry on paraformaldehyde fixed tissues from human, rat, mouse and monkey.

Immunocytochemistry on cells expressing BACE. Acetone or methanol fixation preferred; 4% PFA 5′, RT followed by 0.1% triton X-100 1 hour, can also be used. 1:200-1:500 is recommended, optimization is necessary.

Immunoprecipitation:

Optimal working dilutions must be determined by end user.

Target description

~ 60-75 kDa

Physical form

Format: Purified
Protein A purified
Purified immunoglobulin. Liquid. Buffer = 0.02M Sodium Phosphate, 0.25M NaCl with 0.1% sodium azide.

Storage and Stability

Maintain for 1 year at 2–8°C from date of shipment. Aliquot to avoid repeated freezing and thawing. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
Brain

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Li Zhu et al.
The Journal of biological chemistry, 288(44), 32050-32063 (2013-09-21)
Recent studies link synaptojanin 1 (synj1), the main phosphoinositol (4,5)-biphosphate phosphatase (PI(4,5)P2-degrading enzyme) in the brain and synapses, to Alzheimer disease. Here we report a novel mechanism by which synj1 reversely regulates cellular clearance of amyloid-β (Aβ). Genetic down-regulation of
BACE1 and BACE2 enzymatic activities in Alzheimer's disease.
Ahmed, RR; Holler, CJ; Webb, RL; Li, F; Beckett, TL; Murphy, MP
Journal of Neurochemistry null
Beta-secretase activity increases with aging in human, monkey, and mouse brain.
Fukumoto, H; Rosene, DL; Moss, MB; Raju, S; Hyman, BT; Irizarry, MC
The American Journal of Pathology null
Sindhu Ramesh et al.
PloS one, 13(1), e0190350-e0190350 (2018-01-13)
Honokiol (poly-phenolic lignan from Magnolia grandiflora) is a Sirtuin-3 (SIRT3) activator which exhibit antioxidant activity and augment mitochondrial functions in several experimental models. Modern evidence suggests the critical role of SIRT3 in the progression of several metabolic and neurodegenerative diseases.
Dynamin 1 regulates amyloid generation through modulation of BACE-1.
Zhu, L; Su, M; Lucast, L; Liu, L; Netzer, WJ; Gandy, SE; Cai, D
Testing null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service