Skip to Content
Merck
All Photos(3)

Key Documents

MAB2263

Sigma-Aldrich

Anti-GluR1-NT (NT) Antibody, clone RH95

clone RH95, from mouse

Synonym(s):

AMPA 1, AMPA-selective glutamate receptor 1, Glutamate receptor ionotropic, AMPA 1, glutamate receptor, ionotropic, AMPA 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

RH95, monoclonal

species reactivity

rat

technique(s)

ELISA: suitable
immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... GRIA1(2890)

General description

Glutamate receptors (GluRs) are a diverse group responsible for mediating most of the excitatory synaptic transmission in the CNS of vertebrates. They can be categorized as ionotropic or metabotropic and subcategorized by their agonist preferences (NMDA, AMPA or Kainic acid). There are four types of AMPA selective GluR subunits (GluR1, GluR2, GluR3 and GluR4). Tetrameric or pentameric combinations of different subunits contributes to the functional diversity of AMPA receptors. AMPA receptors mediate fast synaptic current at most excitatory synapses, with stoichiometry characterized by subtype composition. The critical residue controlling calcium permeability is in the pore loop region. In GluR1, GluR3, and GluR4, this position is occupied by a Gln residue. The insertion or removal of GluR1/GluR4 oligomeric channels from postsynaptic membranes appears to be LTP/LTD activity dependent while GluR2/GluR3 oligomers are continuously cycling.

Specificity

This antibody recognizes GluR1 at the N-terminus.

Immunogen

Epitope: N-terminus
Thyroglobulin-conjugated linear peptide of rat GluR1 designed around the N-terminus.

Application

Anti-GluR1-NT (N-terminus) Antibody, clone RH95 is an antibody against GluR1-NT (N-terminus) for use in WB, IP, IC, ELISA.
Research Category
Neuroscience
Research Sub Category
Neurotransmitters & Receptors

Quality

Evaluated by Western Blot in rat brain lysate.

Western Blot Analysis: A 1:1000 dilutions of this antibody detected GluR1 in 10 µg of rat brain lysate.

Target description

110 kDa

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2aκ in buffer containing 0.1 M Tris-Glycine (pH 7.4, 150 mM NaCl) with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Rat brain lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

PKM? knockdown disrupts post-ischemic long-term potentiation via inhibiting postsynaptic expression of aminomethyl phosphonic acid receptors.
Cheng, N; Hu, X; Tian, T; Lu, W
Journal of biomedical research null
The X-linked mental retardation protein OPHN1 interacts with Homer1b/c to control spine endocytic zone positioning and expression of synaptic potentiation.
Nakano-Kobayashi, A; Tai, Y; Nadif Kasri, N; Van Aelst, L
The Journal of Neuroscience null
Paul Farrow et al.
eLife, 4, e09693-e09693 (2015-12-02)
AMPA receptor (AMPAR) function is modulated by auxiliary subunits. Here, we report on three AMPAR interacting proteins-namely CKAMP39, CKAMP52 and CKAMP59-that, together with the previously characterized CKAMP44, constitute a novel family of auxiliary subunits distinct from other families of AMPAR
Jennifer Stanic et al.
Nature communications, 6, 10181-10181 (2015-12-19)
NMDA receptor (NMDAR) composition and synaptic retention represent pivotal features in the physiology and pathology of excitatory synapses. Here, we identify Rabphilin 3A (Rph3A) as a new GluN2A subunit-binding partner. Rph3A is known as a synaptic vesicle-associated protein involved in
Xiaofen Zhong et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 32(37), 12841-12847 (2012-09-14)
MeCP2 (methyl CpG binding protein 2) is a key player in recognizing methylated DNA and interpreting the epigenetic information encoded in different DNA methylation patterns. The functional significance of MeCP2 to the mammalian nervous system is highlighted by the discovery

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service