Skip to Content
Merck
All Photos(2)

Key Documents

HCVD4MAG-67K

Millipore

MILLIPLEX® Human Cardiovascular Disease (CVD) Magnetic Bead Panel 4 - Cardiovascular Disease Multiplex Assay

The analytes available for this multiplex kit are: sE-Selectin, Follistatin, Pecam-1, Pentraxin-3, Tissue Factor (TF), Thrombomodulin, Troponin T (TnT).

Synonym(s):

human CVD immunoassay panel, luminex human cardiovascular disease multiplex assay, millipore human CVD multiplex kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.84

Quality Level

species reactivity

human

manufacturer/tradename

Milliplex®

assay range

accuracy: 103%
(Thrombomodulin)

accuracy: 104%
(Pentraxin-3 (PTX3))

accuracy: 104%
(Tissue Factor (TF))

accuracy: 106%
(Pecam-1)

accuracy: 109%
(sE-Selectin)

standard curve range: 0.01-5 ng/mL
(Troponin T (TnT))

standard curve range: 0.04-30 ng/mL
(Follistatin (FST))

standard curve range: 0.14-100 ng/mL
(Pentraxin-3 (PTX3) & Tissue Factor (TF))

standard curve range: 0.27-200 ng/mL
(Pecam-1)

standard curve range: 0.34-250 ng/mL
(Thrombomodulin)

standard curve range: 2.7-2,000 ng/mL
(sE-Selectin)

inter-assay cv: <20%
intra-assay cv: <10%

technique(s)

multiplexing: suitable

detection method

fluorometric (Luminex xMAP)

shipped in

wet ice

General description

Cardiovascular disease, particularly atherosclerotic vascular disease, is a leading cause of global mortality, accounting for 30% of all global deaths (WHO, 2010). Inflammatory mechanisms play a vital role in the initiation, maintenance, and progression of vascular disease with a strong correlation between inflammatory markers and prognosis in acute and chronic coronary artery disease. Numerous studies have demonstrated an association of obesity and diabetes with cardiovascular risk factors.

MILLIPLEX® Human Cardiovascular Disease (CVD) Panel 4 is an 7-plex kit to be used for the simultaneous quantification of any or all of the following analytes in human serum, plasma or tissue/cell lysate and culture supernatant samples: sE-Selectin, Follistatin (FST), Pecam-1, Pentraxin-3 (PTX-3), Tissue Factor (TF), Thrombomodulin, and Troponin T. This kit uses a 96-well format, contains a lyophilized standard cocktail, two internal assay quality controls and can measure up to 38 samples in duplicate.

The Luminex® xMAP® platform uses a magnetic bead immunoassay format for ideal speed and sensitivity to quantitate multiple analytes simultaneously, dramatically improving productivity while conserving valuable sample volume.

Panel Type: Cardiovascular

Specificity

Cross Reactivty
There was no or negligible cross-reactivity between the antibodies for an analyte and any of the other analytes in this panel.

Application

  • Analytes: sCD31 (sPecam-1), sE-Selectin, Follistatin (FST), Pentraxin-3 (PTX-3), Thrombomodulin, Tissue Factor (TF), Troponin T (TnT)
  • Recommended Sample Type: Human serum, plasma, cell/tissue culture supernatants and lysates
  • Recommended Sample Dilution: 25 μL per well of undiluted serum or plasma; tissue/cell culture samples may require dilution in appropriate control medium
  • Assay Run Time: Overnight (16-18 hours) at 2-8°C
  • Research Category: Cardiovascular Disease
  • Research Subcategory: Metabolic Disorders

Features and Benefits

Design your multiplex kit by choosing available analytes within this panel.

Other Notes

Please contact Technical Service for linearity of dilution.
Sensitivity: See kit protocol for individual analytes sensitive’s.

Legal Information

Luminex is a registered trademark of Luminex Corp
MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
xMAP is a registered trademark of Luminex Corp

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lauren L Ching et al.
Frontiers in pediatrics, 8, 295-295 (2020-07-17)
Kawasaki disease (KD) is the leading cause of acquired pediatric heart disease in the developed world as 25-30% of untreated patients and at least 5% of treated patients will develop irreversible coronary artery lesions (CAL). Pentraxin-3 (PTX-3) has been well-studied
Kévin Contrepois et al.
Cell, 181(5), 1112-1130 (2020-05-30)
Acute physical activity leads to several changes in metabolic, cardiovascular, and immune pathways. Although studies have examined selected changes in these pathways, the system-wide molecular response to an acute bout of exercise has not been fully characterized. We performed longitudinal
Miša Korva et al.
Viruses, 11(8) (2019-07-31)
Hemorrhagic fever with renal syndrome (HFRS) and Crimean-Congo hemorrhagic fever (CCHF) are important viral hemorrhagic fevers (VHF), especially in the Balkan region. Infections with Dobrava or Puumala orthohantavirus and Crimean-Congo hemorrhagic fever orthonairovirus can vary from a mild, nonspecific febrile

Related Content

Multiplex CVD panels like MILLIPLEX® assays enhance biomarker discovery for cardiovascular disease research.

Multiplex CVD panels like MILLIPLEX® assays enhance biomarker discovery for cardiovascular disease research.

Multiplex CVD panels like MILLIPLEX® assays enhance biomarker discovery for cardiovascular disease research.

Multiplex CVD panels like MILLIPLEX® assays enhance biomarker discovery for cardiovascular disease research.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service