Skip to Content
Merck
All Photos(1)

Key Documents

EDSPAK0A1

Milli-Q

EDS-Pak® Polisher

Production of endocrine disruptor-free water at the point of dispense of the Milli-Q® IQ/EQ 7 series

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104212
eCl@ss:
33050190
NACRES:
JA.13

material

carbon cartridge (activated)
polypropylene housing

Quality Level

packaging

pkg of 1 unit

manufacturer/tradename

EDS-Pak®

compatibility

for use with Milli-Q® EQ 7000
for use with Milli-Q® EQ 7008
for use with Milli-Q® EQ 7016
for use with Milli-Q® IQ 7000
for use with Milli-Q® IQ 7003
for use with Milli-Q® IQ 7005
for use with Milli-Q® IQ 7010
for use with Milli-Q® IQ 7015

shipped in

ambient

Looking for similar products? Visit Product Comparison Guide

General description

EDS-Pak® Polisher, an activated carbon-based cartridge, efficiently removes organic contaminants such as phthalates, bisphenol A and nonylphenol. It is validated to produce 300L of endocrine disruptor-free water from Milli-Q® ultrapure water as the water source.
EDS-Pak®-treated ultrapure water is suitable for the preparation of buffers, blanks and standards employed in the quantification of endocrine disruptors.

Application

The EDS-Pak® Polisher is used to produce endocrine disruptor (ED)-free water at the point of dispense of Milli-Q® IQ and EQ 7 series. It efficiently removes trace levels of ED contaminants from ultrapure water for use in laboratory applications, such as to prepare samples, standard solutions or working solutions for the assessment of the environmental toxicity of bisphenols, the analysis of endocrine disrupters in water, the analysis of pharmaceuticals and their degradation products in wastewater, the testing of alkaloids in health food products, and the analysis of alpha-lipoic acid in biological samples by chromatographic techniques.

Features and Benefits

  • Final purification step that delivers endocrine disruptor-free ultrapure water.
  • Validated for efficient removal of bisphenol A, diethylhexyl phthalate, di-n-butyl phthalate and nonylphenol.
  • Certificate of Quality available online.
  • Bottom-tip redesigned for secure installation of the protection bell.
  • Carefree maintenance: Easily installed and replaced.
  • e-Sure tag for RFID connection with Q-POD® dispenser enables full traceability (data management) and automatic consumable status monitoring on the POD’s touchscreen interface.

Legal Information

EDS-PAK is a registered trademark of Merck KGaA, Darmstadt, Germany
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany
Q-POD is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katarzyna Owczarek et al.
Molecules (Basel, Switzerland), 23(12) (2018-12-20)
Bisphenol A (BPA) is one of the most popular and commonly used plasticizer in the industry. Over the past decade, new chemicals that belong to the bisphenol group have increasingly been used in industrial applications as alternatives to BPA. Nevertheless
Teddy Kabeya Kasonga et al.
Data in brief, 25, 104336-104336 (2019-08-28)
In term of pharmaceutical and their intermediate compounds analysis, UPLC/MS method is a valuable equipment to achieve better confirmation on their biodegradation by fungi. The T47D-KBluc reporter gene assay is an appropriate tool to investigate to removal of estrogenic and
Yoshiyuki Kobayashi et al.
Journal of pharmaceutical and biomedical analysis, 166, 435-439 (2019-02-08)
An analytical method for the enantiomeric determination of α-lipoic acid in human urine was developed to evaluate the pharmacokinetics of α-lipoic acid, an ingredient in health food. Urine samples were collected over time after administering α-lipoic acid dietary supplement to

Related Content

Milli-Q® ultrapure water systems ensure reliable science, with an expanded system offer catering to diverse lab needs and constraints.

Milli-Q® ultrapure water systems ensure reliable science, with an expanded system offer catering to diverse lab needs and constraints.

Milli-Q® ultrapure water systems ensure reliable science, with an expanded system offer catering to diverse lab needs and constraints.

Milli-Q® ultrapure water systems ensure reliable science, with an expanded system offer catering to diverse lab needs and constraints.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service