Skip to Content
Merck
All Photos(1)

Key Documents

AB1991

Sigma-Aldrich

Anti-Neurofilament H (200 kDa) Antibody, lysine-serine-proline repeat

serum, Chemicon®

Synonym(s):

Anti-CMT2CC

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

avian, reptile

species reactivity (predicted by homology)

mammals

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... NEFH(4744)

General description

Neurofilaments are a type of intermediate filament that serve as major elements of the cytoskeleton supporting the axon cytoplasm. They are the most abundant fibrillar components of the axon, being on average 3-10 times more frequent than axonal microtubules. Neurofilaments (10nm in dia.) are built from three intertwined protofibrils which are themselves composed of two tetrameric protofilament complexs of monomeric proteins. The neurofilament triplet proteins (68/70, 160, and 200 kDa) occur in both the central and peripheral nervous system and are usually neuron specific. The 68/70 kDa NF-L protein can self-assemble into a filamentous structure, however the 160 kDa NF-M and 200 kDa NF-H proteins require the presence of the 68/70 kDa NF-L protein to co-assemble. Neuromas, ganglioneuromas, gangliogliomas, ganglioneuroblastomas and neuroblastomas stain positively for neurofilaments. Although typically restricted to neurons, neurofilaments have been detected in paragangliomas and adrenal and extra-adrenal pheochromocytomas. Carcinoids, neuroendocrine carcinomas of the skin, and oat cell carcinomas of the lung also express neurofilaments. For more neurofilament information see Nervous System Cell Type Specific Marker chart online under the CHEMICON Technical Support section.

Specificity

Strong reactivity to the major neurofilament subunit HF-H. Since a second neurofilament subunit, NF-M, also contains a few lysine-serine-proline (KSP) sequences, there is generally some reactivity with this protein also. AB1991 stains both phosphorylated and dephosphorylated neurofilaments.

Immunogen

E. Coli recombinant fusion protein containing 37 lysine-serine-proline repeats of rat NF-H.
Epitope: lysine-serine-proline repeat

Application

Detect Neurofilament H (200 kDa) using this Anti-Neurofilament H (200 kDa) Antibody, lysine-serine-proline repeat validated for use in IH(P), WB, IH.
Immunohistochemistry: 1:250-1:1,000 AB1991 is not affected by the level of neurofilament phosphorylation and is particularly good for revealing dendritic and perikaryal neurofilaments.

Immunoblotting

Electron microscopy

Optimal working dilutions must be determined by end user.
Research Category
Neuroscience
Research Sub Category
Neurofilament & Neuron Metabolism

Neuronal & Glial Markers

Physical form

Neat rabbit antisera.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hongyang Jing et al.
Cell & bioscience, 11(1), 105-105 (2021-06-07)
The neuromuscular junction (NMJ) is a peripheral synapse critical to muscle contraction. Like acetylcholine receptors (AChRs), many essential proteins of NMJ are extremely concentrated at the postjunctional membrane. However, the mechanisms of synapse-specific concentration are not well understood; furthermore, it
J A Christianson et al.
Neuroscience, 145(1), 303-313 (2007-01-16)
Human diabetic patients often lose touch and vibratory sensations, but to date, most studies on diabetes-induced sensory nerve degeneration have focused on epidermal C-fibers. Here, we explored the effects of diabetes on cutaneous myelinated fibers in relation to the behavioral
G-Y Xu et al.
Neuroscience, 153(4), 1034-1047 (2008-04-22)
The toxicity of released glutamate contributes substantially to secondary cell death following spinal cord injury (SCI). In this work, the extent and time courses of glutamate-induced losses of neurons and oligodendrocytes are established. Glutamate was administered into the spinal cords
Karra A Muller et al.
Diabetes, 57(6), 1693-1701 (2008-03-26)
Large-fiber diabetic polyneuropathy (DPN) leads to balance and gait abnormalities, placing patients at risk for falls. Large sensory axons innervating muscle spindles provide feedback for balance and gait and, when damaged, can cause altered sensorimotor function. This study aimed to
Ailish Murray et al.
Neural development, 5, 16-16 (2010-06-24)
In the developing hindbrain, cranial motor axon guidance depends on diffusible repellent factors produced by the floor plate. Our previous studies have suggested that candidate molecules for mediating this effect are Slits, Netrin-1 and Semaphorin3A (Sema3A). It is unknown to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service