Skip to Content
Merck
All Photos(1)

Key Documents

8.52417

Sigma-Aldrich

Fmoc-Cys(Dpm)-OH

Novabiochem®

Synonym(s):

Fmoc-Cys(Dpm)-OH, N-α-Fmoc-S-diphenylmethyl-L-cysteine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H27NO4S
CAS Number:
Molecular Weight:
509.62
UNSPSC Code:
12352209
NACRES:
NA.22

Quality Level

product line

Novabiochem®

form

powder

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

thiol

storage temp.

15-25°C

InChI

1S/C31H27NO4S/c33-30(34)28(20-37-29(21-11-3-1-4-12-21)22-13-5-2-6-14-22)32-31(35)36-19-27-25-17-9-7-15-23(25)24-16-8-10-18-26(24)27/h1-18,27-29H,19-20H2,(H,32,35)(H,33,34)/t28-/m0/s1

InChI key

IVPLDYIPIHKRET-NDEPHWFRSA-N

General description

The Dpm group has increased stability compared to Trt, enabling selective removal of Mtt groups on the solid phase with dilute TFA without loss of Dpm groups. Furthermore, the Dpm group is removed by treatment with 95% TFA cleavage cocktails. Racemization during carboxyl activation is lower with this derivative than with Fmoc-Cys(Trt)-OH.

Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Fmoc SPPS
Fmoc SPPS of Cysteine-Containing Peptides

Literature references

[1] H. Hibino, et al. (2014) J. Pept. Sci., 20, 30.
[2] M. Góngora-Benítez, et al. (2012) Org.Lett., 14, 5472.

Analysis Note

Colour (visual): white to off white
Appearance of substance (visual): powder
Identity (IR): passes test
Enantiomeric purity: ≥ 99.5 % (a/a)
Purity (HPLC): ≥ 98.0 % (a/a)
Purity (TLC(011A)): ≥ 98 %
Purity (TLC(0811)): ≥ 98 %
Solubility (1 mmole in 2 ml DMF): clearly soluble
Ethyl acetate (HS-GC): ≤ 0.5 %
Acetate (IC): ≤ 0.1 %

To see the solvent systems used for TLC of Novabiochem® products please click here.

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Overcome challenges in synthesis and disulfide bond formation with protocols for Fmoc solid-phase peptide synthesis of peptides with cysteine and methionine.

Overcome challenges in synthesis and disulfide bond formation with protocols for Fmoc solid-phase peptide synthesis of peptides with cysteine and methionine.

Overcome challenges in synthesis and disulfide bond formation with protocols for Fmoc solid-phase peptide synthesis of peptides with cysteine and methionine.

Overcome challenges in synthesis and disulfide bond formation with protocols for Fmoc solid-phase peptide synthesis of peptides with cysteine and methionine.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service