Skip to Content
MilliporeSigma

Skip To

8.22331

4-Hydroxy-1,3-phenylenediammonium dichloride

for synthesis

Synonym(s):

4-Hydroxy-1,3-phenylenediammonium dichloride, Amidol, 2,4-Diaminophenol dihydrochloride, Phenol-2,4-diammonium dichloride, 1-Hydroxybenzene-2,4-diammonium dichloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 g
Check Cart for Availability
$245.00
250 g
Check Cart for Availability
$602.00

About This Item

Linear Formula:
4-(OH)C6H3-1,3-(NH2)2 · 2 HCl
CAS Number:
Molecular Weight:
197.06
UNSPSC Code:
12352107
EC Index Number:
205-279-4
NACRES:
NA.22
MDL number:
Form:
solid

$245.00


Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

solid

potency

240 mg/kg LD50, oral (Rat)

pH

1-2 (20 °C, 50 g/L in H2O)

mp

205 °C

solubility

275 g/L

bulk density

310 kg/m3

storage temp.

2-30°C

SMILES string

[Cl-].[Cl-].Nc1c(ccc(c1)N)O.[H+].[H+]

InChI

1S/C6H8N2O.2ClH/c7-4-1-2-6(9)5(8)3-4;;/h1-3,9H,7-8H2;2*1H

InChI key

KQEIJFWAXDQUPR-UHFFFAOYSA-N


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1.030678.21102391905
Quality Level

200

Quality Level

100

Quality Level

200

Quality Level

100

solubility

275 g/L

solubility

2000 g/L

solubility

-

solubility

-

storage temp.

2-30°C

storage temp.

2-30°C

storage temp.

2-30°C

storage temp.

-

form

solid

form

solid

form

powder

form

liquid

mp

205 °C

mp

208-212 °C (decomposition)

mp

227-230 °C (decomposition)

mp

-

pH

1-2 (20 °C, 50 g/L in H2O)

pH

1.6 (20 °C, 50 g/L in H2O)

pH

-

pH

-


pictograms

Skull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters