Skip to Content
MilliporeSigma

Skip To

5.38256

IDO Inhibitor

Synonym(s):

IDO Inhibitor, 4-Amino-N-(3-chloro-4-fluorophenyl)-Nʹ-hydroxy-1,2,5-oxadiazole-3-carboximidamide, IDOi, Indoleamine 2,3-Dioxygenase Inhibitor

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
10 mg
Please contact Customer Service for Availability
$267.00

About This Item

Empirical Formula (Hill Notation):
C9H7ClFN5O2
CAS Number:
Molecular Weight:
271.64
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder
Storage condition:
OK to freeze, protect from light

$267.00


Please contact Customer Service for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥98% (HPLC)

Quality Segment

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, protect from light

color

off-white

solubility

DMSO: 50 mg/mL

storage temp.

2-8°C

SMILES string

Fc1c(cc(cc1)\N=C(\NO)/c2n[o]nc2N)Cl

InChI key

HGXSLPIXNPASGZ-UHFFFAOYSA-N

General description

A cell-permeable, bioactive, non-toxic oxadiazolohydroxyamidine based compound that acts as a potent, selective and competitive inhibitor of indoleamine 2,3-dioxygenase (IDO; IC50 = 67 nM for hIDO). Reversibly binds to the catalytically active heme iron (Fe2+) and is inactive towards closely related tryptophan 2,3-dioxygenase (TDO; IC50 >10 µM). Shown to dose-dependently decrease kynurenine levels (IC50 = 19 & 46 nM in HeLa treated with IFN-γ & B16-GM-CSF melanoma cells, respectively) and suppress B16-GM-CSF xenograft tumor growth in C57BL/6 mice (75 mg/kg, s.c., b.i.d.). Though orally bioavailable, but undergoes a rapid clearance (t1/2<0.5 h).

Please note that the molecular weight for this compound is batch-specific due to variable water content. Please refer to the vial label or the certificate of analysis for the batch-specific molecular weight. The molecular weight provided represents the baseline molecular weight without water.

Biochem/physiol Actions

Cell permeable: no
Primary Target
IDO
Reversible: yes
Target IC50: 67 nM for hIDO

Packaging

Packaged under inert gas

Preparation Note

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Yue, E.W., et al. 2009. J. Med. Chem.52, 7364.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
05-840217691444966
assay

≥98% (HPLC)

assay

-

assay

≥95% (HPLC)

assay

≥95% (HPLC)

form

powder

form

-

form

solid

form

solid

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Upstate®

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

storage temp.

2-8°C

storage temp.

-

storage temp.

−20°C

storage temp.

2-8°C

storage condition

OK to freeze, protect from light

storage condition

-

storage condition

OK to freeze, protect from light

storage condition

OK to freeze, protect from light


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters