Skip to Content
Merck
All Photos(1)

Key Documents

476275

Sigma-Aldrich

Myricetin

A cell-permeable flavanoid that displays anti-inflammatory, anti-diabetic and anti-cancer properties.

Synonym(s):

Myricetin, 3,3ʹ,4ʹ,5,5ʹ,7-Hexahydroxyflavone, Hsp70 Inhibitor II

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H10O8
CAS Number:
Molecular Weight:
318.24
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow-brown

solubility

DMSO: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C15H10O8/c16-6-3-7(17)11-10(4-6)23-15(14(22)13(11)21)5-1-8(18)12(20)9(19)2-5/h1-4,16-20,22H

InChI key

IKMDFBPHZNJCSN-UHFFFAOYSA-N

General description

A cell-permeable flavanoid that displays anti-inflammatory, anti-diabetic and anti-cancer properties. Acts as a non-ATP competitive MEK1 inhibitor (IC50<3 µM), and ATP-competitive inhibitors of CKI, CKII, IR, MLCK, PKA and PKC (Ki = 9.0 µM, 0.6 µM, 2.6 µM, 1.7 µM, 27.5 µM and 12.1 µM, respectively) and PI 3-Kα and PIM-1 (IC50 = 1.8 µM and 0.78 µM). Reported to inhibit the activities of phosphodiesterase 1-4 (IC50<40 µM) and MMP-2 (IC50<24 µM in several colorectal carcinoma cells), and block TNFα-mediated NF-κB signaling (at 50 µM in ECV304 cells) and insulin-stimulated glucose transport (Ki = ~ 33.5 µM in rat adipocytes). Further, induce apoptosis in HL-60 leukemia cells with an IC50 of 43 µM.
A cell-permeable flavanoid that displays anti-inflammatory, anti-diabetic and anti-cancer properties. Acts as a non-ATP competitive MEK1 inhibitor (IC50<3 µM), and ATP-competitive inhibitors of CKI, CKII, IR, MLCK, PKA and PKC (Ki = 9.0 µM, 0.6 µM, 2.6 µM, 1.7 µM, 27.5 µM and 12.1 µM, respectively) and PI 3-Kα and PIM-1 (IC50 = 1.8 µM and 0.78 µM). Reported to inhibit the activities of phosphodiesterase 1-4 (IC50<40 µM) and MMP-2 (IC50<24 µM in several colorectal carcinoma cells), and block TNFα-mediated NF-κB signaling (at 50 µM in ECV304 cells) and insulin-stimulated glucose transport (Ki ~ 33.5 µM in rat adipocytes). Further, induce apoptosis in HL-60 leukemia cells with an IC50 of 43 µM.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Lee, K.W., et al. 2007. Carcinogenesis28, 1918.
Holder, S., et al. 2007. Mol. Cancer Ther.6, 163.
Strobel, P., et al. 2005. Biochem. J.386, 471.
Ko, C.H., et al. 2005. Mol. Cancer Ther.4, 281.
Ko, C.H., et al. 2005. Biochem. Pharmacol.69, 913.
Ko, W.C., et al. 2004. Biochem. Pharmacol.68, 2087.
Walker, E.H., et al. 2000. Mol. Cell6, 909.
Agullo, G., et al. 1997. Biochem. Pharmacol.53, 1649.
Hagiwara, M., et al. 1988. Biochem. Pharmacol.37, 2987.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service