Skip to Content
Merck
All Photos(2)

Key Documents

475949

Sigma-Aldrich

NMS-P715

≥98% (HPLC), powder, MPS1 inhibitor, Calbiochem®

Synonym(s):

MPS1 Inhibitor, NMS-P715, (N-(2,6-diethylphenyl)-1-methyl-8-({4-[(1-methylpiperidin-4-yl)carbamoyl]-2-(trifluoromethoxy)phenyl}amino)-4,5-dihydro-1H-pyrazolo[4,3-h]quinazoline-3-carboxamide)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C35H39F3N8O3
CAS Number:
Molecular Weight:
676.73
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

product name

MPS1 Inhibitor, NMS-P715, The MPS1 Inhibitor, NMS-P715 controls the biological activity of MPS1. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Quality Level

Assay

≥98% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 10 mg/mL, clear, colorless

shipped in

ambient

storage temp.

2-8°C

SMILES string

CN(N=C1C(NC2=C(CC)C=CC=C2CC)=O)C3=C1CCC4=C3N=C(NC5=CC=C(C(NC6CCN(C)CC6)=O)C=C5OC(F)(F)F)N=C4

General description

An orally bioavailable, ATP-competitive, pyrazolo-quinazoline, MPS1 inhibitor (IC50 = 182 nM, Ki = 0.99 nM) that is shown to act in a reversible and time-dependent manner. It demonstrates selectivity for MPS1 against a panel of 60 kinases, displaying activity against only three kinases, CK2, MELK, and NEK6 (<10 M), but not against other mitotic kinases including PLK1, CDK1, Aurora A, Aurora B, or the SAC kinase BUB1, in an in vitro kinase assay. It promotes massive SAC (spindle assembly checkpoint) override (EC50 = 65 nM) in nocodazole-arrested U20S cells and elicits a reduction in the G1 and G2/M phase of the cell cycle in A2780 ovarian cancer cells, similar to RNAi-mediated MPS1 silencing. In addition, it is shown to inactivate SAC, delocalize kinetochore components, and inhibit the proliferation of select cancer cell lines (IC50 ~ 1 M), without marked activity among a panel of 127 normal cell lines. Also, it inhibits A2780 tumor xenograft growth in mice (90 mg/kg/day, o.s., in vivo) by 53% without marked body weight loss. Also available as a 5 mM solution in DMSO (Cat. No. 506313).
An orally bioavailable, ATP-competitive, pyrazolo-quinazoline, MPS1 inhibitor (IC50 = 182 nM, Ki = 0.99 nM) that is shown to act in a reversible and time-dependent manner. It demonstrates selectivity for MPS1 against a panel of 60 kinases, displaying activity against only three kinases, CK2, MELK, and NEK6 (<10 µM), but not against other mitotic kinases including PLK1, CDK1, Aurora A, Aurora B, or the SAC kinase BUB1, in an in vitro kinase assay. It promotes massive SAC (spindle assembly checkpoint) override (EC50 = 65 nM) in nocodazole-arrested U20S cells and elicits a reduction in the G1 and G2/M phase of the cell cycle in A2780 ovarian cancer cells, similar to RNAi-mediated MPS1 silencing. In addition, it is shown to inactivate SAC, delocalize kinetochore components, and inhibit the proliferation of select cancer cell lines (IC50 ~ 1 µM), without marked activity among a panel of 127 normal cell lines. Also, it inhibits A2780 tumor xenograft growth in mice (90 mg/kg/day, o.s., in vivo) by 53% without marked body weight loss.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Caldarelli, M., et al. 2011. Bioorg. Med. Chem. Lett.21, 4507.
Colombo, R., et al. 2011. Cancer Res.70, 10255.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shangming Tang et al.
Nature, 606(7916), 930-936 (2022-04-28)
Chromothripsis is a catastrophic mutational process that promotes tumorigenesis and causes congenital disease1-4. Chromothripsis originates from aberrations of nuclei called micronuclei or chromosome bridges5-8. These structures are associated with fragile nuclear envelopes that spontaneously rupture9,10, leading to DNA damage when
Prasad Trivedi et al.
Nature, 618(7967), 1049-1056 (2023-06-15)
Chromothripsis, the shattering and imperfect reassembly of one (or a few) chromosome(s)1, is an ubiquitous2 mutational process generating localized and complex chromosomal rearrangements that drive genome evolution in cancer. Chromothripsis can be initiated by mis-segregation errors in mitosis3,4 or DNA
Akshay Narkar et al.
Cell reports, 34(12), 108892-108892 (2021-03-25)
Most solid tumors are aneuploid, and p53 has been implicated as the guardian of the euploid genome. Previous experiments using human cell lines showed that aneuploidy induction leads to p53 accumulation and p21-mediated G1 cell cycle arrest. We find that
Meenu Maan et al.
Biochimica et biophysica acta. Molecular cell research, 1868(3), 118929-118929 (2020-12-15)
Error-free progression through mitosis is critical for proper cell division and accurate distribution of the genetic material. The anaphase-promoting complex/cyclosome (APC/C) ubiquitin ligase regulates the progression from metaphase to anaphase and its activation is controlled by the cofactors Cdc20 and
Holly Merta et al.
Developmental cell, 56(24), 3364-3379 (2021-12-02)
Failure to reorganize the endoplasmic reticulum (ER) in mitosis results in chromosome missegregation. Here, we show that accurate chromosome segregation in human cells requires cell cycle-regulated ER membrane production. Excess ER membranes increase the viscosity of the mitotic cytoplasm to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service