Skip to Content
Merck
All Photos(1)

Key Documents

440154

Sigma-Aldrich

L-α-lyso-Lecithin, Egg Yolk

Prepared enzymatically from egg yolk lecithin by treatment with phospholipase A.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352211
NACRES:
NA.77

Quality Level

Assay

≥99% (TLC)

form

solid (Free flowing)

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

chloroform: 5 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

Prepared enzymatically from egg yolk lecithin by treatment with phospholipase A.

Biochem/physiol Actions

Cell permeable: no
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Saito, T., et al. 1988. Proc. Natl. Acad. Sci. USA85, 8246.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A method for generating transgenic frog embryos.
E Amaya et al.
Methods in molecular biology (Clifton, N.J.), 97, 393-414 (1999-08-12)
T Saito et al.
Proceedings of the National Academy of Sciences of the United States of America, 85(21), 8246-8250 (1988-11-01)
The effects of lysolecithin (lysophosphatidylcholine) derived from egg yolk as well as of synthetic lysolecithins with different aliphatic chain lengths on tension development of rabbit aortic strips were investigated. Lysolecithins caused slowly progressing, dose-dependent relaxation that was inhibited by hemoglobin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service