Skip to Content
Merck
All Photos(1)

Key Documents

21537

Sigma-Aldrich

IHC Select Secondary Goat Anti-Rabbit IgG Antibody, prediluted, biotinylated

from goat, biotin conjugate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

biological source

goat

Quality Level

conjugate

biotin conjugate

antibody product type

secondary antibodies

clone

polyclonal

species reactivity

rabbit

manufacturer/tradename

Chemicon®
IHC Select

technique(s)

immunohistochemistry: suitable (paraffin)

shipped in

wet ice

target post-translational modification

unmodified

Application

Prediluted for use in Immunohistochemical staining procedures. For example:

Blocking Reagent

Primary Antibody (Rabbit IgG) or Negative Control Reagent

Biotinylated Secondary Antibody (Gt X Rb IgG)

Streptavidin-Enzyme Conjugate

Chromogen

Counterstain

For best results use this reagent in conjunction with other IHC Select detection reagents. A complete list of reagents is available at www.chemicon.com.
This Goat IgG antibody is validated for use in IHC for the detection of the Rabbit IgG protein.

Physical form

Goat Anti-Rabbit IgG in PBS containing carrier protein, with 0.1% Kathon as preservative.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Anke Mittelstädt et al.
Frontiers in immunology, 15, 1253072-1253072 (2024-06-07)
Pancreatic ductal adenocarcinoma (PDAC) is one of the deadliest forms of cancer and peritoneal dissemination is one major cause for this poor prognosis. Exosomes have emerged as promising biomarkers for gastrointestinal cancers and can be found in all kinds of
Rui Zheng et al.
Molecular medicine reports, 22(3), 2351-2363 (2020-07-25)
One key risk factor of aortic valve stenosis in clinical practice is bicuspid aortic valve (BAV). Increasing evidence indicates that numerous microRNAs (miRs/miRNAs) are involved in BAV calcification via their target genes. miR‑330‑3p was found to be involved in the deterioration
Stéphane Louryan et al.
Journal of morphology, 283(2), 207-218 (2021-12-16)
Parrot embryos carry peculiar appendages at their developing beak that have been described as pseudoteeth. To better characterize the pattern of development responsible for the emergence of these dental appendages, we examined parrot embryos combining conventional histology and microtomography approaches.
Gang Wang et al.
Oncology letters, 20(6), 389-389 (2020-11-17)
Colorectal cancer (CRC) is the third most common malignant type of tumor worldwide. Neurensin-2 (NRSN2) is a small neuronal membrane protein associated with tumorigenesis. Therefore, the present study aimed to investigate the association between NRSN2 and CRC, and further examined
J T Martin et al.
Scientific reports, 7(1), 15765-15765 (2017-11-19)
Total disc replacement with an engineered substitute is a promising avenue for treating advanced intervertebral disc disease. Toward this goal, we developed cell-seeded disc-like angle ply structures (DAPS) and showed through in vitro studies that these constructs mature to match

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service