Skip to Content
Merck
All Photos(1)

Documents

178615

Sigma-Aldrich

Anti-Aquaporin 5 Rabbit pAb

liquid, Calbiochem®

Synonym(s):

Anti-AQP5

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

liquid

does not contain

preservative

species reactivity

rat, mouse, human

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

Anti-Aquaporin 5, rabbit polyclonal, recognizes the ~28 kDa AQP 5 protein in epithelial tissue extracts. It is validated for ELISA, Western blotting, immunofluorescence, and immunohistochemistry.
Immunoaffinity purified rabbit polyclonal antibody. Recognizes the ~28 kDa AQP5 protein.
Recognizes the ~28 kDa AQP 5 protein in epithelial tissue extracts.

Immunogen

a synthetic peptide corresponding to a 17-amino acid sequence in the cytoplasmic region of rat AQP5

Application

ELISA (0.5-1.0 µg/ml)

Immunoblotting (1-10 µg/ml)

Immunofluorescence (2-20 µg/ml)

Immunohistochemistry (2-20 µg/ml)

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Analysis Note

Positive Control
Epithelial cells

Other Notes

Does not cross-react with other AQP proteins. The use of 0.5-1.0% milk in all primary/secondary antibody-enzyme conjugate incubations is recommended in order to suppress non-specific bands. Variables associated with assay conditions will dictate the optimal working dilution.
Lee, M.D., et al. 1996. J. Biol. Chem. 271, 8599.
Raina, S., et al. 1995. J. Biol. Chem. 270, 1908.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Cristina Kenney et al.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society, 52(10), 1341-1350 (2004-09-24)
Corneas with edema-related diseases lose transparency, which causes significant vision loss. This study analyzed seven aquaporins (AQPs) in normal corneas, pseudophakic/aphakic bullous keratopathy (PBK/ABK) corneas, Fuchs' dystrophy corneas, keratoconus corneas, post-cataract surgery (PCS) corneas, and normal organ-cultured corneas. RNA levels
Amy P Wong et al.
The Journal of clinical investigation, 119(2), 336-348 (2009-01-24)
The bone marrow compartment is enriched in stem and progenitor cells, and an unidentified subpopulation of these cells can contribute to lung epithelial repair. Here we identify this subpopulation and quantitate its relative contribution to injured airway epithelium. A subpopulation
Lucas T Woods et al.
PloS one, 10(5), e0123641-e0123641 (2015-05-09)
Transforming growth factor-β (TGF-β) is a multi-functional cytokine with a well-described role in the regulation of tissue fibrosis and regeneration in the liver, kidney and lung. Submandibular gland (SMG) duct ligation and subsequent deligation in rodents is a classical model
Devendra Kumar Maurya et al.
PloS one, 10(6), e0129171-e0129171 (2015-06-13)
TMEM16A/ANO1 is a calcium-activated chloride channel expressed in several types of epithelia and involved in various physiological processes, including proliferation and development. During mouse embryonic development, the expression of TMEM16A in the olfactory epithelium is dynamic. TMEM16A is expressed at
Fernanda de Paula et al.
Journal of anatomy, 238(3), 794-806 (2020-10-21)
Aquaporins (AQPs) are essential to coordinate the transit of water and ions through the cell membrane. In salivary glands (SGs), AQPs have been associated with saliva formation, facilitating water absorption through the epithelium during the formation of hypotonic saliva, which

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service