Skip to Content
Merck
All Photos(1)

Key Documents

1.50629

Supelco

Purospher®STAR RP-8 endcapped (2 µm) HPLC Columns

L × I.D. 100 mm x 2.1 mm Hibar® HR UHPLC Column

Synonym(s):

Purospher® STAR RP-8 endcapped (2µm) Hibar® HR 100-2.1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
NACRES:
SB.52

product name

Purospher® STAR RP-8 endcapped (2 µm), L × I.D. 100 mm x 2.1 mm Hibar® HR UHPLC column

material

stainless steel column

Quality Level

Agency

suitable for USP L7

product line

Purospher®

feature

endcapped

packaging

1 ea of

extent of labeling

11.2% Carbon loading

parameter

1000 bar pressure
65 °C max. temp.

technique(s)

LC/MS: suitable
UHPLC-MS: suitable
UHPLC: suitable
mass spectrometry (MS): suitable (High)

column L × I.D.

10 cm × 2.1 mm

surface area

330 m2/g

matrix

fully porous particle

matrix active group

C8 bonding phase

particle size

2 μm

pore size

120 Å pore size

operating pH range

1.5-10.5

separation technique

reversed phase
supercritical fluid (SFC)

storage temp.

no temp limit

General description

Purospher® STAR RP-8 endcapped, 2 µm UHPLC columns are designed for use in UHPLC Applications and instruments providing full UHPLC resolution and highest separation efficiency′s at maximum pressure of 1000 bar.Purospher® STAR RP-8 endcapped, 2 µm UHPLC columns allow the separation of basic, neutral, and metal chelating compounds with simple mobile phases and excellent peak symmetry. They are based on ultra-pure fully porous silica particles (Type B). These columns offer an outstanding stability from pH 1.5 to 10.5 over a wide temperature range. Purospher® STAR RP-8 endcapped, 2 µm Hibar® HR 100-2.1 is a ready to use UHPLC column suitable for connection to any UHPLC system providing a pressure stability of 1000 bar.The column dimension of 100 mm length x 2.1 mm I.D. is the optimal choice for fast UHPLC separation of complex samples allowing significant solvent savings and improved sensitivity.Learn more about Purospher® STAR HPLC and UHPLC ColumnsLearn more about the Supelco HPLC Column portfolio and Method development:Brochure: HPLC and UHPLC Column Selection GuideBrochure: A Practical Guide to HPLC - Method Development Guide

Application

Intended to be used in the separation of aflatoxins, aldehydes, alkaloids, antibiotics, amino acids, esters, fatty acids, ketones, nitrosamines, nucleosides, PCBs, peptides, phenols, preservatives, steroids, and metabolized steroids.

Features and Benefits

  • Ultra-pure silica (99.999%) for excellent peak symmetry
  • High separation efficiency
  • Reproducible results from run-to-run and batch-to-batch
  • Outstanding pH stability from pH 1.5 to 10.5
  • Good suitability for LC-MS applications

Legal Information

Hibar is a registered trademark of Merck KGaA, Darmstadt, Germany
Purospher is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service