Skip to Content
Merck
All Photos(1)

Key Documents

1.44196

Millipore

Rotor

for use with RCS® High Flow Touch

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122409
NACRES:
NA.74

material

anodized aluminum

Agency

non-compendial

feature

autoclavable

packaging

pkg of 1 kit

application(s)

air monitoring
cosmetics
food and beverages
pharmaceutical

compatibility

for use with RCS® High Flow Touch

shipped in

ambient

storage temp.

room temp

General description

Rotor is an additional accessory for use with RCS® microbial air sampler. The RCS® portable air sampler is used for fast and easy monitoring of ambient air and compressed gases in controlled environments. The microbial air sampler is based on impaction by centrifugal force. The Rotor is made of autoclavable aniodized aluminum and is used to accelerate the incoming air. Each combination of an RCS® Air Sampler and rotor must be calibrated separately.

Application

Rotor is an additional accessory with the RCS® microbial air sampler used to accelerate the incoming air. These air samplers find application in microbial monitoring of ambient air in pharmaceutical, medical and food industry.

Compatibility

The Rotor is compatible for use with RCS® Air Samplers of the High Flow series (RCS® High Flow Touch, RCS® High Flow) and RCS® Isolator.

Legal Information

RCS is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Ensure microbial air sampling generates precise results complying with international standards. Our systems are easy-to-use and reliable.

Ensure microbial air sampling generates precise results complying with international standards. Our systems are easy-to-use and reliable.

Ensure microbial air sampling generates precise results complying with international standards. Our systems are easy-to-use and reliable.

Ensure microbial air sampling generates precise results complying with international standards. Our systems are easy-to-use and reliable.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service