Skip to Content
Merck
All Photos(2)

Key Documents

14-1067

Sigma-Aldrich

Human Recombinant MeCP2 (methyl-CpG-binding protein 2)

Human recombinant N-terminal His-tagged MeCP2 expressed in SF9 insect cells & purified by Fast Protein Liquid Chromatography (FPLC) & demonstrated to bind methylated DNA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.32

recombinant

expressed in Sf9 cells

Quality Level

Assay

>95% (SDS-PAGE)

mol wt

Mw 50.2 kDa (including histidine tag)

concentration

0.5 μg/μL

shipped in

dry ice

General description

Human recombinant N-terminal His-tagged MeCP2 expressed in SF9 insect cells and purified by Fast Protein Liquid Chromatography (FPLC).
MeCP2 (methyl-CpG-binding protein 2) binds methylated CpG sequences in double-stranded DNA resulting in the suppression of transcription. This protein is likely to be involved in the repression of multiple genes. Consistent with this role, mutations of MeCP2 are linked to Rett syndrome, a progressive neurological disorder that is one of the most common causes of mental retardation in females.

Application

Research Category
Epigenetics & Nuclear Function

Physical form

In a buffer containing 20 mM Tris-Cl pH 7.6, 1 mM EDTA, 0.15 M NaCl, 10% glycerol, 0.5 mM PMSF and 1 mM DTT.

Storage and Stability

Stable at -80ºC for 6 months from date of receipt.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service