Skip to Content
Merck
All Photos(1)

Documents

12666

Sigma-Aldrich

Albumin, Human Serum, Fraction V, Low Heavy Metals

Synonym(s):

Albumin, Human Serum, Fraction V, Low Heavy Metals

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.25

Assay

≥95% (SDS-PAGE)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

impurities

≤5.0% Ash
≤6% moisture

pI 

4.7

solubility

water: 50 mg/mL

cation traces

heavy metals: ≤20 ppm

shipped in

ambient

storage temp.

2-8°C

InChI

1S/C3F8/c4-1(5,2(6,7)8)3(9,10)11

InChI key

QYSGYZVSCZSLHT-UHFFFAOYSA-N

General description

Human serum albumin (HSA) is an abundant protein found in the plasma. This globular, soluble, and unglycosylated monomeric protein, has three homologous domains (I, II, and III). These domains have subdomains A and B having common structural motifs. The HSA gene is located on the human chromosome 4q13.3.

Application

Albumin, Human Serum, Fraction V, Low Heavy Metals has been used:
  • as a supplement in double buffer and milliQ-water for in vivo
  • in serum-free media for polarized secretion assays in human dermal microvascular endothelial cells
  • as a supplement in sodium phosphate buffer for the reconstitution of hormone lysis samples

Biochem/physiol Actions

Human serum albumin (HSA) is very crucial in stabilizing extracellular fluid volume. It functions as an excipient for vaccines and therapeutic protein drugs. HSA serves as a cell culture medium supplement for the synthesis of vaccines and pharmaceuticals. It is useful as a carrier protein for steroids, fatty acids, and thyroid hormones. HSA is used to treat cardiopulmonary bypass, trauma, hemorrhage, and hypoalbuminemia. HSA is useful in biological applications, such as implantable biomaterials, biochromatography, surgical adhesives, and sealants.

Warning

Toxicity: Standard Handling (A)

Preparation Note

Prepared from serum that has been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tina Jorsal et al.
The Journal of clinical endocrinology and metabolism, 104(12), 6403-6416 (2019-07-06)
After Roux-en-Y gastric bypass (RYGB) surgery, postprandial plasma glucagon concentrations have been reported to increase. This occurs despite concomitant improved glucose tolerance and increased circulating plasma concentrations of insulin and the glucagon-inhibiting hormone glucagon-like peptide 1 (GLP-1). To investigate whether
Chanchal Sadhu et al.
Journal of immunology (Baltimore, Md. : 1950), 170(5), 2647-2654 (2003-02-21)
Neutrophil chemotaxis is a critical component of the innate immune response. Neutrophils can sense an extremely shallow gradient of chemoattractants and produce relatively robust chemotactic behavior. This directional migration requires cell polarization with actin polymerization occurring predominantly in the leading
Samuel A J Trammell et al.
Molecules (Basel, Switzerland), 25(10) (2020-05-24)
Farnesoid X receptor (FXR) and Takeda G-protein coupled receptor 5 (TGR5) are the two known bile acid (BA) sensitive receptors and are expressed in the intestine and liver as well as in extra-enterohepatic tissues. The physiological effects of extra-enterohepatic FXR/TRG5
Nicolai J Wewer Albrechtsen et al.
American journal of physiology. Endocrinology and metabolism, 311(2), E302-E309 (2016-06-02)
Glucagon is a metabolically important hormone, but many aspects of its physiology remain obscure, because glucagon secretion is difficult to measure in mice and rats due to methodological inadequacies. Here, we introduce and validate a low-volume, enzyme-linked immunosorbent glucagon assay
C J Kelley et al.
Microbiology spectrum, 10(6), e0360122-e0360122 (2022-11-08)
As multidrug and pan-resistance among Enterobacterales continue to increase, there is an urgent need for more therapeutic options to treat these infections. New β-lactam and β-lactam inhibitor (BLI) combinations have a broad spectrum of activity, but those currently approved do

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service