Skip to Content
MilliporeSigma

1.16996

pH Test Strips, reflectometric

pH range 4.0-9.0, REFLECTOQUANT®, for use with RQflex® 20

Synonym(s):

pH chemical test strips, pH test strip for instrument reader

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41116116
NACRES:
NB.21
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

pH Test, reflectometric, pH range 4.0-9.0

feature

non-bleeding

Quality Segment

pH range

4.0-9.0

application(s)

agriculture
aquaculture
beverage testing
drinking water
food testing
ground and surface water
mineral water
swimming pools
wastewater

compatibility

RQflex® 20

detection method

reflectometric

storage temp.

15-25°C

General description

In water analysis the pH value plays a very important role in how other substances behave. The test can be used for monitoring the pH value in aqueous solutions, beverages, e.g. wine, after dilution and food after sample pretreatment. The Reflectoquant pH tests are suited for all media in environmental analysis and in industrial in-process controls. The reaction zones of the test strip change color depending on the pH. The responsible indicator is chemically bound to the zones and hence does not bleed. The resulting color is evaluated reflectometrically on the RQflex® 20. For tests for other parameters that require a specific pH range of the sample (e.g. Spectroquant, see instructions of use), we recommend the use of Reflectoquant or the semi-quantitative MQuant pH test strips as pre-test.
Method: reflectometric with test strips pH 4.0 - 9.0 Reflectoquant®

Analysis Note

function control: conforms
pH-value
- standard 1:
3.8 - 4.2
- standard 2: 6.8 - 7.2
- standard 3: 8.8 - 9.2

Legal Information

REFLECTOQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany
RQFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany


Storage Class

11 - Combustible Solids

wgk

WGK 1



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library