Skip to Content
Merck
All Photos(1)

Key Documents

1.14897

Supelco

Chloride Test, photometric

photometric, 2.5-250 mg/L (Cl-), Spectroquant®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

product name

Chloride Test, photometric, 2.5-250 mg/L (Cl-), Spectroquant®

product line

Spectroquant®

Quality Level

usage

sufficient for 100 tests
sufficient for 175 tests (depending on package size)

specific analyte(s)

chloride

measuring range

2.5-250 mg/L (Cl-)

technique(s)

photometry: suitable

compatibility

for use with Spectroquant® Move 100
for use with Spectroquant® Nova 60 A
for use with Spectroquant® Prove 100
for use with Spectroquant® Prove 300
for use with Spectroquant® Prove 600

detection method

photometric (Iron(III)-thiocyanate)

storage temp.

15-25°C

General description

Method: photometric 2.5 - 250 mg/l Cl⁻ Spectroquant®
This Spectroquant® Chloride test allows the accurate quantification of chloride ions in various waters and waste water. Their concentration depends on geological factors and the general local situation. Within the test chloride ions react with mercury(II)-thiocyanate to mercury(II) chloride. The liberated thiocyanide reacts with iron ions to form a read complex. The method is analogous to EPA 325.1 and APHA 4500-Cl- E.
All our Cell and Reagent Test Kits are equipped with the unique Live ID (2D barcode) which allows seamless method recognition and contains essential information such as lot number, expiry date, and automatic calibration updates.
The Spectroquant® Reagent Test Kits contain highly stable, ready-to-use reagent mixtures to perform the analysis according to the procedure described in the accompanying instruction leaflet.

Application

  • PREFUL MRI for Monitoring Perfusion and Ventilation Changes after Elexacaftor-Tezacaftor-Ivacaftor Therapy for Cystic Fibrosis: A Feasibility Study.: This study demonstrates the use of PREFUL MRI to monitor changes in perfusion and ventilation in cystic fibrosis patients post-therapy, showcasing its potential for non-invasive assessment in clinical settings (Dohna et al., 2024).
  • Wearable sweat chloride sensors: materials, fabrication and their applications.: Reviews advances in materials and fabrication techniques for wearable sweat chloride sensors, discussing their significant potential for real-time health monitoring and diagnostics (Patel et al., 2024).
  • Initial characteristics of cystic fibrosis in Algeria: Description of 34 pediatric cases.: Provides an overview of the clinical presentation and initial characteristics of cystic fibrosis in a pediatric Algerian cohort, contributing to the regional understanding of this genetic disorder (Bendoukha et al., 2024).

Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - STOT RE 2

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

188.6 °F

Flash Point(C)

87 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Luis A Cueto et al.
Environmental science and pollution research international, 27(14), 15984-15993 (2019-05-06)
Phosphorus (P) is a nutrient for plant growth but also a pollutant in water bodies causing eutrophication. The source of P is mainly human and animal wastewater and runoffs from different land uses. The objective of the present study is

Related Content

Clean drinking water is essential for food and beverage production. This article gives an overview of chemical and microbiological testing of the drinking water for quality and safety.

Clean drinking water is essential for food and beverage production. This article gives an overview of chemical and microbiological testing of the drinking water for quality and safety.

Clean drinking water is essential for food and beverage production. This article gives an overview of chemical and microbiological testing of the drinking water for quality and safety.

Clean drinking water is essential for food and beverage production. This article gives an overview of chemical and microbiological testing of the drinking water for quality and safety.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service