Skip to Content
Merck
All Photos(1)

Key Documents

1.11962

Supelco

Azomethine H

GR for analysis (reagent for the determination of boron)

Synonym(s):

Azomethine H

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H12NNaO8S2
CAS Number:
Molecular Weight:
445.40
UNSPSC Code:
12352107
EC Index Number:
227-698-1
NACRES:
NA.21

Quality Level

form

solid

bulk density

500 kg/m3

storage temp.

15-25°C

InChI

1S/C17H13NO8S2.Na.H2O/c19-15-4-2-1-3-10(15)9-18-14-7-12(27(21,22)23)5-11-6-13(28(24,25)26)8-16(20)17(11)14;;/h1-9,18,20H,(H,21,22,23)(H,24,25,26);;1H2/q;+1;/p-1/b10-9+;;

InChI key

BEDIAJJRAYEWBI-TTWKNDKESA-M

Application


  • Ethanol and Higher Alcohols′ Production in Fungal and Bacterial Laboratory Cultures and Significance for Forensic Samples.: This study investigates the microbial production of ethanol and higher alcohols including 1-Propanol, highlighting their roles in forensic science for identifying post-mortem biochemical changes (Boumba et al., 2024).

  • Excess Enthalpies Analysis of Biofuel Components: Sunflower Oil-Alcohols Systems.: This study provides a thermodynamic analysis of biofuel mixtures involving 1-Propanol, optimizing energy content and stability in renewable energy resources (Golikova et al., 2024).


Analysis Note

Identity (IR-spectrum): conforms
Spec. Absorptivity A 1%/1cm (236 nm; 0.05 %; 1 cm; buffer solution pH 5.1): ≥ 1200
Loss on Drying (105°C): ≤ 2.0 %
Suitability for photometric (determination of boron): conforms

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service