Skip to Content
Merck
All Photos(1)

Key Documents

1.04967

Supelco

Potassium cyanide

for analysis EMSURE® ACS,ISO,Reag. Ph Eur

Synonym(s):

Potassium cyanide, Cyanogen potassium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CKN
CAS Number:
Molecular Weight:
65.12
MDL number:
UNSPSC Code:
12352302
EC Index Number:
205-792-3
NACRES:
NA.21

grade

ACS reagent

Quality Level

Agency

reag. ISO
reag. Ph. Eur.

product line

EMSURE®

Assay

≥97% (silver nitrate titration)

form

solid

potency

7.49 mg/kg LD50, oral (Rat)
14.29 mg/kg LD50, skin (Rabbit)

pH

11-12 (20 °C, 20 g/L in H2O)

bp

1625 °C/1013 hPa

mp

634 °C

density

1.55 g/cm3 at 20 °C

bulk density

750 kg/m3

anion traces

chloride (Cl-): ≤0.02%
phosphate (PO43-): ≤0.005%
sulfate (SO42-): ≤0.01%
sulfide (S2-): ≤0.0005%
thiocyanate (SCN-): ≤0.01%

cation traces

Fe: ≤0.01%
Na: ≤0.5%
Pb: ≤0.0002%

storage temp.

no temp limit

InChI

1S/CN.K/c1-2;/q-1;+1

InChI key

NNFCIKHAZHQZJG-UHFFFAOYSA-N

Application


  • Identification of Iminium Intermediates Generation in the Metabolism of Tepotinib Using LC-MS/MS: In Silico and Practical Approaches to Bioactivation Pathway Elucidation.: The research identifies iminium intermediates in the metabolism of tepotinib using liquid chromatography-mass spectrometry. Potassium cyanide is utilized to trap reactive intermediates, providing insights into drug metabolism pathways (Abdelhameed et al., 2020).

  • Synthesis of stable-isotope-labeled N-(3-dimethylaminopropyl)-N′-ethylcarbodiimide and N-(3-dimethylaminopropyl)-N′-ethylurea.: This paper describes the synthesis of stable-isotope-labeled compounds using potassium cyanide in the synthesis process to understand the chemical structure and reactivity of these labeled compounds (Cao et al., 2020).

  • Carbon-13 synthesis and NMR spectroscopic geometric isomer evaluation to support the filing of teriflunomide.: The study involves the synthesis of carbon-13 labeled teriflunomide and its isomers, using potassium cyanide in the synthetic pathway. The NMR spectroscopic analysis helped in the evaluation of geometric isomers (Kurz et al., 2021).

  • A hybrid lipid membrane coating "shape-locks" silver nanoparticles to prevent surface oxidation and silver ion dissolution.: This research develops a hybrid lipid membrane to stabilize silver nanoparticles, using potassium cyanide in the analytical phase to measure the prevention of silver ion dissolution and oxidation (Miesen et al., 2020).

Linkage

Replaces: PX1435; PX1435-5; PX1435-1; PX1435-20

Analysis Note

Assay (argentometric): ≥ 97.0 %
Chloride (Cl): ≤ 0.02 %
Phosphate (PO₄): ≤ 0.005 %
Sulfate (SO₄): ≤ 0.01 %
Sulphide (S): ≤ 0.0005 %
Thiocyanate (SCN): ≤ 0.01 %
Fe (Iron): ≤ 0.01 %
Na (Sodium): ≤ 0.5 %
Pb (Lead): ≤ 0.0002 %
Corresponds to ACS,Reag. Ph Eur

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Oral - Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Met. Corr. 1 - STOT RE 1

Target Organs

Thyroid

Supplementary Hazards

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service