Skip to Content
Merck
All Photos(1)

Key Documents

1.00496

Sigma-Aldrich

Formaldehyde solution 4%, buffered, pH 6.9

(approx. 10% Formalin solution) for histology

Synonym(s):

Formaline solution, Methanal solution, Methyl aldehyde solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116124
NACRES:
NA.21
application(s):
diagnostic assay manufacturing
hematology
histology
bp:
100 °C/1013 hPa
Pricing and availability is not currently available.

Quality Level

form

liquid

IVD

for in vitro diagnostic use

concentration

3.8-4.2% (acidimetric)

color

colorless

pH

6.7-7.1 ( in H2O)

bp

100 °C/1013 hPa

mp

-92 °C

density

1.017 g/cm3 at 20 °C

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

15-25°C

General description

Formaldehyde (HCOH) is one of the most familiar and effective fixatives. Formaldehyde fixation is suitable for nearly all types of histological tissue, as well as calcified deposits, bone, cartilage, nervous system specimens. The fixation of the histological specimen material of human or animal origin is commonly performed in a 4% or 10% Formaldehyde solution. Chemically, the 4 % Formaldehyde solution equals a 10 % Formalin solution. This Formaldehyde solution 4% is a ready-to-use working solution that is stabilized with phosphate buffer to protect tissue structures. The development of formic acid will be effective prevented. It is an IVD registered and CE certified product and can be applied for diagnostical purposes. For more details, please see instructions for use (IFU). The IFU can be downloaded from this webpage.

Pictograms

Exclamation markHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 1B - Muta. 2 - Skin Sens. 1

Storage Class Code

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Soham Gupta et al.
Frontiers in immunology, 11, 437-437 (2020-04-01)
The hijacking of cellular function through expression of proteins that interfere with the activity of cellular enzymes and regulatory complexes is a common strategy used by viruses to remodel the cell environment in favor of their own replication and spread.

Protocols

How to stain organoids? A complete step-by-step protocol for immunofluorescent (IF) and immunocytochemical (ICC) staining of organoid cultures using antibodies

How to stain organoids? A complete step-by-step protocol for immunofluorescent (IF) and immunocytochemical (ICC) staining of organoid cultures using antibodies

How to stain organoids? A complete step-by-step protocol for immunofluorescent (IF) and immunocytochemical (ICC) staining of organoid cultures using antibodies

How to stain organoids? A complete step-by-step protocol for immunofluorescent (IF) and immunocytochemical (ICC) staining of organoid cultures using antibodies

See All

Related Content

Learn about the criticality of biological tissue staining for research and clinical pathology using standard and special stains and dyes.

Learn about the criticality of biological tissue staining for research and clinical pathology using standard and special stains and dyes.

Questions

1–2 of 2 Questions  
  1. Is it okay to fix an entire tissue with 4% PFA at 37°C overnight using product 1004960700 without altering its morphology?

    1 answer
    1. The temperature used for standard tissue processing historically has been room temperature. Temperature can affect the rate of fixation, and modern tissue processors may use elevated temperatures to reduce processing time without adverse effects on tissue morphology. Cold fixation is common in electron microscopy when using glutaraldehyde as a fixative.

      Helpful?

  2. Do 4% formaldehyde or 10% formalin solutions contain traces of methanol?

    1 answer
    1. Product 1004960700 and all other formaldehyde or formalin solutions contain methanol to prevent the formaldehyde from reforming as a polymer or escaping as a gas. If a methanol-free solution is required, paraformaldehyde powders or crystals can be purchased and prepared into methanol-free solutions, although these solutions are unstable and should be prepared fresh daily or kept for only short periods of time.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service