Skip to Content
Merck
All Photos(1)

Key Documents

1.00304

Supelco

Hydrobromic acid

47%, EMPLURA®

Synonym(s):

Hydrogen bromide solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352301
NACRES:
NA.21

product name

Hydrobromic acid 47%, EMPLURA®

vapor pressure

10.6 hPa ( 20 °C)

Quality Level

product line

EMPLURA®

Assay

≥47% (acidimetric)

form

liquid

impurities

≤0.0005% Fe (Iron)
≤0.001% Heavy metals (as Pb)
≤0.002% Phosphate, Phosphite (as PO4)
≤0.005% Sulfate and sulphite (as SO4)
≤0.01% Non volatile matter
≤0.05% Chloride (Cl)

bp

126 °C/1013 hPa

mp

-11 °C

density

1.49 g/cm3 at 20 °C

storage temp.

2-30°C

Application


  • The use of ion chromatography for the determination of volatile inorganic acids in workplace air: Hydrobromic acid is used to validate methodology for air quality assessment in workplaces, ensuring safety and compliance with environmental health standards. This application underscores the essential nature of 47% hydrobromic acid in maintaining chemical safety in industrial environments (Kowalska and Szewczyńska, 2022).

Analysis Note

Assay (acidimetric): ≥ 47 %
Chloride (Cl): ≤ 0.05 %
Phosphate, Phosphite (as PO₄): ≤ 0.002 %
Sulfate and sulphite (as SO₄): ≤ 0.005 %
Heavy metals (as Pb): ≤ 0.001 %
Fe (Iron): ≤ 0.0005 %
Non volatile matter: ≤ 0.01 %

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service