Skip to Content
Merck
All Photos(2)

Key Documents

05-1470

Sigma-Aldrich

Anti-Raptor Antibody, clone 1H6.2

clone 1H6.2, from mouse

Synonym(s):

P150 target of rapamycin (TOR)-scaffold protein, p150 target of rapamycin (TOR)-scaffold protein containing WD-repeats, regulatory associated protein of mTOR

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1H6.2, monoclonal

species reactivity

mouse, human

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... RPTOR(57521)
mouse ... Rptor(74370)

General description

Raptor is a subunit of mTORC1 which regulates cell growth and survival in response to nutrient and hormonal signals. mTORC1 is activated in response to growth factors or amino-acids. Within mTORC1, Raptor positively regulates mTORC1 activity and functions as a scaffold for recruiting mTORC1 substrates.

Specificity

This antibody recognizes Raptor.

Immunogen

Epitope: Unknown
Recombinant protein corresponding to human Raptor.

Application

Anti-Raptor Antibody, clone 1H6.2 detects level of Raptor & has been published & validated for use in WB, IP & IC.
Immunocytochemistry Analysis: A 1:500 dilution from a previous lot detected Raptor in NIH/3T3 cells.

Immunoprecipitation Analysis: 20 µg from a previous lot immunoprecipitated Raptor in HEK293 cell lysate.
Research Category
Signaling
Research Sub Category
PI3K, Akt, & mTOR Signaling

Quality

Evaluated by Western Blot in HEK293 cell lysate.

Western Blot Analysis: 1 µg/ml of this antibody detected Raptor on 10 µg of HEK 293 cell lysate.

Target description

~ 150 kDa

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4, 150 mM NaCl) with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HEK 293 cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Julia Dyachok et al.
The Journal of biological chemistry, 291(43), 22414-22426 (2016-09-03)
The mechanistic target of rapamycin complex 1 (mTORC1) coordinates cell growth with its nutritional, hormonal, energy, and stress status. Amino acids are critical regulators of mTORC1 that permit other inputs to mTORC1 activity. However, the roles of individual amino acids
Abdessattar Khlaifia et al.
Molecular brain, 15(1), 56-56 (2022-06-18)
Hippocampal CA1 parvalbumin-expressing interneurons (PV INs) play a central role in controlling principal cell activity and orchestrating network oscillations. PV INs receive excitatory inputs from CA3 Schaffer collaterals and local CA1 pyramidal cells, and they provide perisomatic inhibition. Schaffer collateral
Milagros Suarez et al.
Oncotarget, 12(10), 955-966 (2021-05-21)
The MAPK-interacting kinases 1 and 2 (MNK1/2) have generated increasing interest as therapeutic targets for acute myeloid leukemia (AML). We evaluated the therapeutic potential of the highly-selective MNK1/2 inhibitor Tomivosertib on AML cells. Tomivosertib was highly effective at blocking eIF4E
Radhika Chadha et al.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology, 45(6), 1059-1067 (2020-01-18)
Abnormal neurotransmission is central to schizophrenia (SZ). Alterations across multiple neurotransmitter systems in SZ suggest that this illness may be associated with dysregulation of core intracellular processes such as signaling pathways that underlie the regulation and integration of these systems.
Shannon McNulty et al.
Journal of virology, 87(2), 912-922 (2012-11-09)
Hantavirus pulmonary syndrome (HPS) is a severe respiratory disease characterized by pulmonary edema, with fatality rates of 35 to 45%. Disease occurs following infection with pathogenic New World hantaviruses, such as Andes virus (ANDV), which targets lung microvascular endothelial cells.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service