Skip to Content
Merck
All Photos(1)

Documents

D-927

Supelco

Doxepin hydrochloride solution

1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H22ClNO
CAS Number:
Molecular Weight:
315.84
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

CN(C)CC/C=C1C2=C(C=CC=C2)COC3=C/1C=CC=C3.[H]Cl

InChI

1S/C19H21NO.ClH/c1-20(2)13-7-11-17-16-9-4-3-8-15(16)14-21-19-12-6-5-10-18(17)19;/h3-6,8-12H,7,13-14H2,1-2H3;1H/b17-11-;

InChI key

MHNSPTUQQIYJOT-CULRIWENSA-N

General description

Doxepin is a tricyclic antidepressant used for the treatment of sleep maintenance and sold under trade names such as Silenor® and Aponal. This analytical reference standard is appropriate for use as starting material in calibrators or controls for numerous LC/MS or GC/MS applications such as forensic analysis, clinical toxicology, urine drug testing, or pharmaceutical research.

Application


  • Magnetic Particle Spray Mass Spectrometry: This study utilizes advanced analytical techniques, potentially incorporating Doxepin hydrochloride as a standard in mass spectrometry, to improve detection sensitivities for antidepressants and other neuropharmacological agents (Venga Mendes and Figueiredo, 2024).

  • Desipramine induces anti-inflammatory dorsal root ganglion transcriptional signatures: Though primarily about desipramine, the methodologies and findings could be paralleled in studies using Doxepin hydrochloride to understand its effects on neuroinflammation and pain management (Serafini et al., 2024).

  • Effect of lipid emulsion on neuropsychiatric drug-induced toxicity: Reviews the mitigation of toxicity from drugs like Doxepin hydrochloride, underscoring the importance of lipid emulsions in treating drug overdoses and adverse effects (Hwang and Sohn, 2024).

  • Novel Ti surface coated with PVA hydrogel and chitosan nanoparticles with antibacterial drug release: While focused on surface coatings, the methodology could be adapted for controlled release studies of tricyclic antidepressants like Doxepin hydrochloride in biomedical applications (Carrazco Ávila et al., 2024).

  • Doxepin is more effective than zolpidem in improving executive function in patients with insomnia disorder: This clinical study on Doxepin could relate directly to Doxepin hydrochloride′s use in sleep medicine and psychiatric research, illustrating its efficacy and therapeutic potential (Yu et al., 2023).



Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Silenor is a registered trademark of Somaxon Pharmaceuticals, Inc.
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adam Wichniak et al.
Polski merkuriusz lekarski : organ Polskiego Towarzystwa Lekarskiego, 31(181), 65-70 (2011-08-30)
Disturbed sleep is a core symptom of depression and is among diagnostic criteria for depressive episode. Effects of an antidepressant drug on sleep are important for its clinical profile. Rapid improvement of sleep quality is particularly indicated in depressed patients
Yuliya Levintova et al.
International journal of pharmaceutics, 419(1-2), 96-106 (2011-08-02)
When liquid donors/receivers are used for in vitro skin permeation studies, excess hydration can change skin properties compared to in vivo conditions. A novel in vitro method of determining the permeability of drugs through skin was developed that avoids exposing
Yunes Panahi et al.
Skinmed, 9(3), 152-158 (2011-06-17)
Oral doxepin was shown to reduce chronic pruritus due to sulfur mustard. The present study compared the effects of topical doxepin 5% with betamethasone 1% for the treatment of pruritus in veterans exposed to sulfur mustard. This investigator-blinded, randomized, clinical
Donald R Lynch et al.
Cardiology journal, 19(4), 434-438 (2012-07-25)
Vorinostat is a histone deacetylase inhibitor used in the treatment of recurrent or persistent cases of cutaneous T-cell lymphoma (CTCL). A retrospective review of 116 patients from phase I and II clinical trials who had a baseline and at least
Suppression of extrapyramidal side effects of doxepin by thalamic deep brain stimulation for Tourette syndrome.
L Rzesnitzek et al.
Neurology, 77(18), 1708-1709 (2011-11-02)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service