Skip to Content
Merck

Skip To

B-034

Bupropion hydrochloride solution

1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C13H19Cl2NO
CAS Number:
Molecular Weight:
276.20
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
EC Number:
200-659-6
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

certified reference material

Quality Segment

description

Drug Control: Kábítószer / Narcotic Drug (Hungary), 78/2022. (XII. 28.) BM rendelet

form

liquid

feature

SNAP-N-SPIKE®, SNAP-N-SHOOT®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Kábítószer / Narcotic Drug (Hungary), 78/2022. (XII. 28.) BM rendelet

concentration

1.0 mg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

Cl.CC(NC(C)(C)C)C(=O)c1cccc(Cl)c1

InChI

1S/C13H18ClNO.ClH/c1-9(15-13(2,3)4)12(16)10-6-5-7-11(14)8-10;/h5-9,15H,1-4H3;1H

InChI key

HEYVINCGKDONRU-UHFFFAOYSA-N

Gene Information

General description

A certified solution standard suitable for use as starting material in calibrators or controls for a variety of LC/MS or GC/MS applications from forensic analysis and clinical toxicology to urine drug testing and pharmaceutical research. Bupropion is an antidepressant and smoking cessation drug sold under the trade names Wellbutrin® and Zyban®.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany
Wellbutrin is a registered trademark of GlaxoSmithKline LLC
Zyban is a registered trademark of Everris International B.V.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
B-052H-066C-118
technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

technique(s)

gas chromatography (GC): suitable, liquid chromatography (LC): suitable

format

single component solution

format

single component solution

format

single component solution

format

single component solution

form

liquid

form

liquid

form

liquid

form

liquid

Quality Level

300

Quality Level

300

Quality Level

300

Quality Level

300

grade

certified reference material

grade

certified reference material

grade

certified reference material

grade

certified reference material

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C


signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

target_organs

Eyes,Central nervous system

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

49.5 °F - closed cup

flash_point_c

9.7 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Rapid assessment of cardiovascular risk among users of smoking cessation drugs within the US Food and Drug Administration's Mini-Sentinel program.
Sengwee Toh et al.
JAMA internal medicine, 173(9), 817-819 (2013-03-27)
Georgia Tsiapa et al.
In vivo (Athens, Greece), 27(2), 171-176 (2013-02-21)
Smoking is related to a great variety of pathological conditions, many of which affect the respiratory system, such as lung cancer and chronic obstructive pulmonary disease (COPD). Smoking cessation should be an integral part of the therapeutic approach to patients
Marney A White et al.
The Journal of clinical psychiatry, 74(4), 400-406 (2013-05-10)
Binge-eating disorder (BED) is defined by recurrent binge eating (eating unusually large quantities of food during which a subjective loss of control is experienced), marked distress about the binge eating, and the absence of inappropriate weight compensatory behaviors. BED is



Global Trade Item Number

SKUGTIN
B-034-1ML04061833419137

Questions

Reviews

No rating value

Active Filters