Skip to Content
Merck
All Photos(1)

Key Documents

900406P

Avanti

pacFA GalCer

Avanti Research - A Croda Brand 900406P, powder

Synonym(s):

D-galactosyl-β-1,1′ N-(9-(3-pent-4-ynyl-3-H-diazirin-3-yl)-nonanoyl)-D-erythro-sphingosine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C39H69N3O8
CAS Number:
Molecular Weight:
707.98
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 1 mg (900406P-1mg)
pkg of 1 × 5 mg (900406P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 900406P

lipid type

click reagents

shipped in

dry ice

storage temp.

−20°C

SMILES string

C#CCCCC1(N=N1)CCCCCCCCC(N[C@](CO[C@H]2[C@H](O)[C@@H](O)[C@@H](O)[C@@H](CO)O2)([H])[C@H](O)/C=C/CCCCCCCCCCCCC)=O

General description

The pacFA lipid contains a photoactivable diazirine ring with a clickable alkyne group.

Biochem/physiol Actions

D-galactosyl-β-1,1′ N-(9-(3-pent-4-ynyl-3-H-diazirin-3-yl)-nonanoyl)-D-erythro-sphingosine (pacFA GalCer) may be used for a global analysis of protein-lipid interactions in living cells. pacFA acts as a precursor for the biosynthesis of bifunctional lipids. Proteins in contact with the bifunctional lipids are then cross-linked by ultraviolet (UV) irradiation of the diazirine ring. Finally, click chemistry is used to label the alkyne with a reporter molecule.

Packaging

5 mL Amber Glass Screw Cap Vial (900406P-1mg)
5 mL Amber Glass Screw Cap Vial (900406P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Per Haberkant et al.
Biochimica et biophysica acta, 1841(8), 1022-1030 (2014-01-21)
Understanding biological processes at the mechanistic level requires a systematic charting of the physical and functional links between all cellular components. While protein-protein and protein-nucleic acid networks have been subject to many global surveys, other critical cellular components such as
In vivo profiling and visualization of cellular protein-lipid interactions using bifunctional fatty acids.
Per Haberkant et al.
Angewandte Chemie (International ed. in English), 52(14), 4033-4038 (2013-03-02)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service