Skip to Content
Merck
All Photos(1)

Key Documents

860073W

Avanti

C18:0 GM3-d5 (synthetic)

Avanti Research - A Croda Brand 860073W, methanol solution

Synonym(s):

C18:0 GM3 Ganglioside-d5 (synthetic, ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C59H106D5N3O21
CAS Number:
Molecular Weight:
1203.55
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

methanol solution

packaging

pkg of 1 × 1 mL (860073W-100ug)

manufacturer/tradename

Avanti Research - A Croda Brand 860073W

concentration

0.1 mg/mL (860073W-100ug)

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@](/C=C/CCCCCCCCCCCCC)(O)[C@@]([H])(NC(CCCCCCCCCCCCCCCC([2H])(C([2H])([2H])[2H])[2H])=O)CO[C@H]1[C@H](O)[C@@H](O)[C@H](O[C@@H]2O[C@H](CO)[C@@H]([C@H](O[C@@]3(C([O-])=O)O[C@@H]([C@H](O)[C@H](O)CO)[C@@](NC(C)=O)([H])[C@@H](O)C3)[C@H]2O)O)[C@@H](CO)O1.[

General description

C18:0 GM3-d5 is a deuterated monosialodihexosylganglioside (GM3) ganglioside wherein the terminal ethyl (-CH2-CH3) protons of stearic acid are replaced by deuterium.

Packaging

2 mL Amber Glass Sealed Ampule (860073W-100ug)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Magali Pettazzoni et al.
PloS one, 12(7), e0181700-e0181700 (2017-07-28)
The biological diagnosis of sphingolipidoses currently relies on the measurement of specific enzymatic activities and/or genetic studies. Lysosphingolipids have recently emerged as potential biomarkers of sphingolipidoses and Niemann-Pick type C in plasma. We developed a sensitive and specific method enabling

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service