Skip to Content
Merck
All Photos(3)

Key Documents

W280523

Sigma-Aldrich

1-Octen-3-ol

greener alternative

natural, ≥95%, FG

Synonym(s):

Pentyl vinyl carbinol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4CH(OH)CH=CH2
CAS Number:
Molecular Weight:
128.21
FEMA Number:
2805
Beilstein:
1744110
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
2.023
NACRES:
NA.21

grade

FG
Fragrance grade
Halal
Kosher
natural

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 172.515

Assay

≥95%

greener alternative product characteristics

Less Hazardous Chemical Syntheses
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

refractive index

n20/D 1.437 (lit.)

bp

84-85 °C/25 mmHg (lit.)

density

0.837 g/mL at 20 °C
0.83 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

greener alternative category

Organoleptic

mushroom; musty; earthy

SMILES string

CCCCCC(O)C=C

InChI

1S/C8H16O/c1-3-5-6-7-8(9)4-2/h4,8-9H,2-3,5-7H2,1H3

InChI key

VSMOENVRRABVKN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Octen-3-ol occurs naturally in mushrooms. It is the most prominent odor volatile produced by fungi such as Aspergillus, Penicillium and Fusarium species. 1-Octen-3-ol is also formed in major quantity when soybeans are soaked in water as a pretreatment before soymilk production. It is also a potent mosquito attractant.
We are committed to bringing you Greener Alternative Products, which adhere to one of the four categories of Greener Alternatives . This product is a Biobased products, showing key improvements in Green Chemistry Principles “Less Hazardous Chemical Syntheses” and “Use of Renewable Feedstock”.

Application


  • Fluctuation of flavor quality in roasted duck: The consequences of raw duck preform′s repetitive freeze-thawing.: This study explores how repetitive freeze-thaw cycles of raw duck preforms influence the flavor quality of roasted duck, examining specific volatile compounds including 1-Octen-3-ol and its impact on sensory attributes (Gao et al., 2024).

  • A new HS-SPME-GC-MS analytical method to identify and quantify compounds responsible for changes in the volatile profile in five types of meat products during aerobic storage at 4 °C.: This research introduces a novel analytical approach using HS-SPME-GC-MS to track changes in volatile profiles, including the role of 1-Octen-3-ol, across different meat products under specific storage conditions (Acquaticci et al., 2024).

  • Comprehensive investigation on the dynamic changes of volatile metabolites in fresh scent green tea during processing by GC-E-Nose, GC-MS, and GC × GC-TOFMS.: This detailed study assesses how volatile metabolites like 1-Octen-3-ol evolve during the processing of fresh scent green tea, utilizing advanced gas chromatography techniques (Wang et al., 2024).

  • Insights into "wheat aroma": Analysis of volatile components in wheat grains cultivated in saline-alkali soil.: This article examines the ′wheat aroma′ by analyzing volatile components, including 1-Octen-3-ol, in wheat grains grown in challenging saline-alkali soils, highlighting the adaptative traits of these crops (Sun et al., 2024).

  • Demonstrating the Applicability of Proton Transfer Reaction Mass Spectrometry to Quantify Volatiles Emitted by the Mycoparasitic Fungus Trichoderma atroviride in Real Time: Monitoring of Trichoderma-Based Biopesticides.: This study demonstrates the use of proton transfer reaction mass spectrometry to quantify volatile organic compounds, including 1-Octen-3-ol, emitted in real-time by the fungus Trichoderma atroviride, used in biopesticide applications (Lochmann et al., 2024).


Other Notes

Natural occurrence: Clover, cooked artichoke, fish, mushroom, peas, soybeans, wheat bread, tea, and beef.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

154.4 °F - closed cup

Flash Point(C)

68 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Studies on mushroom flavours 3. Some flavour compounds in fresh, canned and dried edible mushrooms.
Dijkstra FY.
Zeitschrift fur Lebensmittel-Untersuchung Und -Forschung, 160(4), 401-405 (1976)
Carbon dioxide and 1-octen-3-ol as mosquito attractants.
Takken W & Kline DL.
Journal of the American Mosquito Control Association, 5(3), 311-316 (1989)
D L Kline
Journal of the American Mosquito Control Association, 10(2 Pt 2), 280-287 (1994-06-01)
The role of octenol as a mosquito attractant is still under investigation. When used alone, octenol has been a good attractant for only a few species. However, there appears to be a synergistic response of species of the genera Aedes
Volatile flavor compounds produced by molds of Aspergillus, Penicillium, and Fungi imperfecti.
Kaminski E, et al.
Applied Microbiology, 27(6), 1001-1004 (1974)
The formation of 1-octen-3-ol in soybeans during soaking.
Badenhop AF & Wilkens WF.
Journal of the American Oil Chemists' Society, 46(3), 179-182 (1969)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service