Skip to Content
Merck
All Photos(1)

Key Documents

N20204

Sigma-Aldrich

4-Nitrophenylacetic acid

ReagentPlus®, 99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H4CH2CO2H
CAS Number:
Molecular Weight:
181.15
Beilstein:
1911801
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

product line

ReagentPlus®

Assay

99%

form

powder

mp

150-155 °C (lit.)

SMILES string

OC(=O)Cc1ccc(cc1)[N+]([O-])=O

InChI

1S/C8H7NO4/c10-8(11)5-6-1-3-7(4-2-6)9(12)13/h1-4H,5H2,(H,10,11)

InChI key

YBADLXQNJCMBKR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shao-ting Wang et al.
Guang pu xue yu guang pu fen xi = Guang pu, 22(3), 412-413 (2003-08-27)
The binary solid complexes had been synthesized using rare-earth ion with p-nitrophenylacetic acid. The chemical formula of ReL3.H2O(Re = Eu, Sm, Tb) complexes was determined by elemental analysis, molar conductivity, and TG-DTA. The composition structure and properties of the binary
Y L Yang et al.
Guang pu xue yu guang pu fen xi = Guang pu, 21(5), 680-681 (2003-08-30)
The europium ternary solid complex containing p-nitrophenylacetic acid and 1, 10-phenanthroline was synthesized in this work, and the chemical formula of this compound was determined to be EuL3 phen by elemental analysis. The complex has been characterized by analysis of
F Liao et al.
Clinica chimica acta; international journal of clinical chemistry, 314(1-2), 67-76 (2001-11-24)
Conventional enzyme activities make use of the initial reaction rate at high substrate concentrations. Because this is not always practical, alternative enzyme assays have been sought. Reaction curve fitting with an integrated rate equation was investigated to assay serum arylesterase
D Yang et al.
Toxicological sciences : an official journal of the Society of Toxicology, 64(1), 20-27 (2001-10-19)
Carboxylesterases play an important role in the metabolism of endogenous lipids and foreign compounds; therefore, xenobiotic regulation of carboxylesterase gene expression has both physiological and pharmacological significance. We previously reported that beta-naphthoflavone and 3-methylcholanthrene, two potent inducers for cytochrome P4501A
Halis T Balaydın et al.
European journal of medicinal chemistry, 54, 423-428 (2012-06-13)
A series of bisphenol, bromophenol, and methoxyphenol derivatives (2-24) including the natural bromophenols vidalol B, 3,4,6-tribromo-5-(2,5-dibromo-3,4-dihydroxybenzyl)benzene-1,2-diol (2) and 5,5'-methylenebis(3,4,6-tribromo-benzene-1,2-diol) (3) were prepared. In the current study, inhibition of four human carbonic anhydrase (hCA, EC 4.2.1.1) isozymes, I, II, IV, and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service