Skip to Content
Merck
All Photos(4)

Key Documents

M9138

Sigma-Aldrich

L-(−)-Malic acid disodium salt

≥95% (titration)

Synonym(s):

L-Hydroxybutanedioic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C4H4O5Na2
CAS Number:
Molecular Weight:
178.05
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95% (titration)

form

powder

SMILES string

[Na].O[C@@H](CC(O)=O)C(O)=O

InChI

1S/C4H6O5.2Na/c5-2(4(8)9)1-3(6)7;;/h2,5H,1H2,(H,6,7)(H,8,9);;/q;2*+1/p-2/t2-;;/m0../s1

InChI key

WPUMTJGUQUYPIV-JIZZDEOASA-L

Application

L-(−)-Malic acid disodium salt is a disodium salt of L-malic acid that is used as a resolving agent to separate enantiomers of certain racemic compounds. It is also used as a chiral auxiliary in organic synthesis for the preparation of a wide range of organic compounds.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Duoduo Chen et al.
Molecular microbiology, 87(6), 1237-1244 (2013-01-26)
Nicotine is an environmental toxicant in tobacco waste, imposing a serious hazard for human health. Some bacteria including Pseudomonas spp. strains are able to metabolize nicotine to non-toxic compounds. The pyrrolidine pathway of nicotine degradation in Pseudomonas putida S16 has
Euan Burton et al.
Canadian journal of microbiology, 58(12), 1378-1388 (2012-12-06)
Clostridium thermocellum is a Gram-positive thermophilic anaerobic bacterium with the ability to directly convert cellulosic biomass into useful products such as ethanol and hydrogen. In this study, a quantitative comparative proteomic analysis of the organism was performed to identify proteins
Cuiyue Liang et al.
Plant physiology, 161(3), 1347-1361 (2013-01-24)
Low pH, aluminum (Al) toxicity, and low phosphorus (P) often coexist and are heterogeneously distributed in acid soils. To date, the underlying mechanisms of crop adaptation to these multiple factors on acid soils remain poorly understood. In this study, we
E González-Fandos et al.
Poultry science, 92(7), 1936-1941 (2013-06-19)
This work evaluated the effect of malic acid washing on the growth of Listeria monocytogenes on poultry legs stored at 4°C for 8 d. Fresh inoculated chicken legs were dipped into a 1 or 2% malic acid solution (vol/vol) for
Frank Norbert Gellerich et al.
IUBMB life, 65(3), 180-190 (2013-02-13)
This review focuses on problems of the intracellular regulation of mitochondrial function in the brain via the (i) supply of mitochondria with ADP by means of ADP shuttles and channels and (ii) the Ca(2+) control of mitochondrial substrate supply. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service