Skip to Content
Merck
All Photos(1)

Key Documents

GF30053540

T1 - thermocouple alloy, Ni90Cr10

foil, 2m coil, 38mm coil width, 0.60mm thickness, annealed

Synonym(s):

NI090300

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
11101711
NACRES:
NA.23

form

annealed
foil (coil)

manufacturer/tradename

Goodfellow 300-535-40

L × W × thickness

2 m × 38 mm × 0.60 mm

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service